Skip to Content
Merck
All Photos(1)

Key Documents

SML0349

Sigma-Aldrich

Neferine

≥98% (HPLC)

Synonym(s):

(-)-Neferine, 4′′-O-Methylliensinine, 4-[[(1S)-1,2,3,4-Tetrahydro-6,7-dimethoxy-2-methyl-1-isoquinolinyl]methyl]-2-[[(1R)-1,2,3,4-tetrahydro-6-methoxy-1-[(4-methoxyphenyl)methyl]-2-methyl-7-isoquinolinyl]oxy]-phenol, Methylliensinine, Neferin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H44N2O6
CAS Number:
Molecular Weight:
624.77
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -34 to -44° (c=0 3. CHCl3)

storage condition

desiccated
protect from light

color

white to beige

solubility

DMSO: >15 mg/mL

storage temp.

2-8°C

SMILES string

COc1ccc(C[C@H]2N(C)CCc3cc(OC)c(Oc4cc(C[C@@H]5N(C)CCc6cc(OC)c(OC)cc56)ccc4O)cc23)cc1

InChI

1S/C38H44N2O6/c1-39-15-14-27-21-36(44-5)38(23-30(27)31(39)17-24-7-10-28(42-3)11-8-24)46-34-19-25(9-12-33(34)41)18-32-29-22-37(45-6)35(43-4)20-26(29)13-16-40(32)2/h7-12,19-23,31-32,41H,13-18H2,1-6H3/t31-,32+/m1/s1

InChI key

MIBATSHDJRIUJK-ZWXJPIIXSA-N

Application

Neferine has been used to study its effect in autophagy signalling.

Biochem/physiol Actions

Neferine is a bis-benzylisoquinoline alkaloid isolated from the green seed embryos Nelumbo Nucifera Gaertn (Lotus) that displays multiple pharmacological activities including anti-tumor, antidepressant-like and antiarrhythmic actions. Recent studies shown that neferine potently inhibits proliferation of human osteosarcoma cells while is low-toxic on non-neoplastic human osteoblast cells. Apparently in U2OS cells, neferine stabilizes p21 through the activation of p38 MAPK, which leads to G1 cell cycle arrest. Also, neferine inhibits Ang II-induced HUVSMC proliferation by upregulation of HO-1.

Features and Benefits

This compound is featured on the MAPKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong-fang Gu et al.
Naunyn-Schmiedeberg's archives of pharmacology, 380(2), 143-151 (2009-05-09)
We studied the effects of isoquinoline alkaloid neferine (Nef) extracted from the seed embryo of Nelumbo nucifera Gaertn on Human ether-à-go-go-related gene (HERG) channels stably expressed in human embryonic kidney (HEK293) cells using whole-cell patch clamp technique, western blot analysis
Yong Zhao et al.
Phytotherapy research : PTR, 26(2), 277-283 (2011-06-17)
The purpose of this study was to investigate the in vitro inhibition potential of the three purified herbal constituents tetrahydropalmatine (Tet), neferine (Nef) and berberine (Ber) towards recombinant human CYP1A2, CYP2D6 and CYP3A4 metabolic activities. In vitro incubations were performed
Sadik Altan Ozal et al.
Cutaneous and ocular toxicology, 39(2), 97-105 (2020-02-18)
Purpose: Proliferative vitreoretinopathy (PVR) occurs in approximately 5-10% of patients after retinal detachment surgery. Neferine is a bis-benzylisoquinoline alkaloid found in the green seed embryos (Nelumbo nucifera) of the lotus flower and has various properties, such as being antithrombotic, antioxidant
Shao Liu et al.
Journal of separation science, 32(14), 2476-2481 (2009-06-27)
A preparative high-speed counter-current chromatography method for separation and purification of liensinine, isoliensinine and neferine from seed embryo of Nelumbo nucifera GAERTN was successfully established by using n-hexane-ethyl acetate-methanol-water (5:8:4:5, v/v, containing 0.5% NH(4)OH) as the two-phase solvent system. From
Yumi Sugimoto et al.
European journal of pharmacology, 634(1-3), 62-67 (2010-02-24)
The effects of neferine, an alkaloid of Nelumbo nucifera Gaertner embryos, on immobility in the forced swimming test, which is used to evaluate antidepressants, were investigated in mice. The administration of neferine from 25 to 100 mg/kg i.p. elicited anti-immobility

Articles

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service