Skip to Content
Merck
All Photos(1)

Key Documents

SML1428

Sigma-Aldrich

O4I2

≥98% (HPLC)

Synonym(s):

2-[(4-Chlorophenyl)amino]-4-thiazolecarboxylic acid ethyl ester, Ethyl 2-((4-chlorophenyl)amino)thiazole-4-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$394.00

$394.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$394.00

About This Item

Empirical Formula (Hill Notation):
C12H11ClN2O2S
CAS Number:
Molecular Weight:
282.75
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$394.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(OCC)C1=CSC(NC2=CC=C(Cl)C=C2)=N1

InChI

1S/C12H11ClN2O2S/c1-2-17-11(16)10-7-18-12(15-10)14-9-5-3-8(13)4-6-9/h3-7H,2H2,1H3,(H,14,15)

InChI key

ULUBAPWNHROTEU-UHFFFAOYSA-N

Biochem/physiol Actions

O4I2 is a potent inducer of the Octamer-binding transcription factor 4 (Oct3/4) in various human cell lines including human fibroblasts. In principle O4I2 could be a useful tool for chemical-mediated induction of pluripotency in somatic cells.
Potent inducer of Oct3/4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service