Skip to Content
Merck
All Photos(1)

Documents

SML0651

Sigma-Aldrich

OAC1

≥98% (HPLC)

Synonym(s):

N-1H-Pyrrolo[2,3-c]pyridin-5-yl-benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11N3O
CAS Number:
Molecular Weight:
237.26
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C14H11N3O/c18-14(10-4-2-1-3-5-10)17-13-8-11-6-7-15-12(11)9-16-13/h1-9,15H,(H,16,17,18)

InChI key

HWJRIFZDXJKJJN-UHFFFAOYSA-N

Biochem/physiol Actions

OAC1 enhances iPSC reprogramming efficiency by enhancing Oct4 and Nanog driven gene expression. Treatment of mouse embryonic fibroblasts transduced with Oct2, Sox2, Klf4 and c-myc displayed a two-fold increase in the formation of ESC-like clones. OAC1 treated cells display a hypomethylated state of Oct4 promoter DNA.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service