Skip to Content
Merck
All Photos(1)

Key Documents

SML1371

Sigma-Aldrich

Prucalopride

≥98% (HPLC)

Synonym(s):

4-Amino-5-chloro-2,3-dihydro-N-[1-(3-methoxypropyl)-4-piperidinyl]-7-benzofurancarboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$90.70
50 MG
$352.00

$90.70


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$90.70
50 MG
$352.00

About This Item

Empirical Formula (Hill Notation):
C18H26ClN3O3
CAS Number:
Molecular Weight:
367.87
UNSPSC Code:
12352200
NACRES:
NA.77

$90.70


Estimated to ship onMay 26, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

COCCCN1CCC(NC(C2=CC(Cl)=C(N)C3=C2OCC3)=O)CC1

InChI

1S/C18H26ClN3O3/c1-24-9-2-6-22-7-3-12(4-8-22)21-18(23)14-11-15(19)16(20)13-5-10-25-17(13)14/h11-12H,2-10,20H2,1H3,(H,21,23)

InChI key

ZPMNHBXQOOVQJL-UHFFFAOYSA-N

Biochem/physiol Actions

In healthy male individuals, prucalopride decreases the display of esophageal acid and promotes gastric emptying.[1] It is effective, safe and shows good tolerance for the treatment of men with chronic constipation.[2]
Prucalopride is a selective 5-HT4 serotonin receptor agonist with enterokinetic activity.
Prucalopride is a selective 5-HT4 serotonin receptor agonist with enterokinetic activity. 5-HT4 receptors are GPCRs that are expressed in the CNS and peripheral tissues. The receptor has a role in GI motility disorders such as irritable bowel syndrome and idiopathic constipation. Agonists are prokinetic and of potential use for disoroders of reduced motility. Prucalopride is a selective 5-HT4 serotonin receptor agonist with enterokinetic activities. Prucalopride has pK(i) values of 8.60 and 8.10 for the human 5-HT(4a) and 5-HT(4b) receptor, respectively, and at least 290-fold selectivity for 5-HT4 over other serotonin and dopamine receptors.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Randomized, Double-Blind, Placebo-Controlled, Phase 3 Trial to Evaluate the Efficacy, Safety, and Tolerability of Prucalopride in Men With Chronic Constipation.
Yiannakou Y, et al.
The American Journal of Gastroenterology, 110(5), 741-741 (2015)
Prucalopride decreases esophageal acid exposure and accelerates gastric emptying in healthy subjects.
Kessing BF, et al.
Neurogastroenterology and Motility, 26(8), 1079-1086 (2014)
Wael Noor El-Nachef et al.
Development (Cambridge, England), 147(13) (2020-06-17)
The enteric nervous system (ENS) is essential for normal gastrointestinal function. Although the embryonic origin of enteric neurons from the neural crest is well established, conflicting evidence exists regarding postnatal enteric neurogenesis. Here, we address this by examining the origin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service