M2946
Mosapride citrate salt dihydrate
≥98% (HPLC), solid, 5-HT₄ receptor agonist
Synonym(s):
4-Amino-5-chloro-2-ethoxy-N-[[4-[(4-fluorophenyl)methyl ]-2-morpholinyl]methyl]benzamide citrate dihydrate
About This Item
Recommended Products
Product Name
Mosapride citrate salt dihydrate,
Assay
≥98% (HPLC)
form
solid
solubility
DMSO: ≥5 mg/mL
originator
Takeda
storage temp.
2-8°C
SMILES string
O.O.OC(=O)CC(O)(CC(O)=O)C(O)=O.CCOc1cc(N)c(Cl)cc1C(=O)NCC2CN(CCO2)Cc3ccc(F)cc3
InChI
1S/C21H25ClFN3O3.C6H8O7.2H2O/c1-2-28-20-10-19(24)18(22)9-17(20)21(27)25-11-16-13-26(7-8-29-16)12-14-3-5-15(23)6-4-14;7-3(8)1-6(13,5(11)12)2-4(9)10;;/h3-6,9-10,16H,2,7-8,11-13,24H2,1H3,(H,25,27);13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);2*1H2
InChI key
KVKIQHMTGSGTFO-UHFFFAOYSA-N
Biochem/physiol Actions
Features and Benefits
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
We offer many products related to serotonin receptors for your research needs.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service