Skip to Content
Merck
All Photos(1)

Key Documents

SML2739

Sigma-Aldrich

SM1-71-R

≥98% (HPLC)

Synonym(s):

N-[2-[[5-Chloro-2-[[4-(4-methyl-1-piperazinyl)phenyl]amino]-4-pyrimidinyl]amino]phenyl]propanamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$98.10
25 MG
$397.00

$98.10


Estimated to ship onApril 03, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$98.10
25 MG
$397.00

About This Item

Empirical Formula (Hill Notation):
C24H28ClN7O
CAS Number:
Molecular Weight:
465.98
UNSPSC Code:
12352200
NACRES:
NA.77

$98.10


Estimated to ship onApril 03, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Clc1c(nc(nc1)Nc3ccc(cc3)N4CCN(CC4)C)Nc2c(cccc2)NC(=O)CC

InChI

1S/C24H28ClN7O/c1-3-22(33)28-20-6-4-5-7-21(20)29-23-19(25)16-26-24(30-23)27-17-8-10-18(11-9-17)32-14-12-31(2)13-15-32/h4-11,16H,3,12-15H2,1-2H3,(H,28,33)(H2,26,27,29,30)

InChI key

IIMJCIGRKJCCHT-UHFFFAOYSA-N

Biochem/physiol Actions

SM1-71-R, a reversible analog of multi-targeted kinase covalent inhibitor SM1-71, is a cell penetrant and potent multi-targeted kinase inhibitor. SM1-71-R can be used as a washout control to distinguish between covalent and non-covalent inhibition.
cell penetrant and potent multi-targeted kinase inhibitor that can be used as a washout control for covalent inhibitor SM1-71

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suman Rao et al.
The Journal of biological chemistry, 294(21), 8664-8673 (2019-03-13)
Most cancer cells are dependent on a network of deregulated signaling pathways for survival and are insensitive, or rapidly evolve resistance, to selective inhibitors aimed at a single target. For these reasons, drugs that target more than one protein (polypharmacology)
Li Tan et al.
Bioorganic & medicinal chemistry, 25(3), 838-846 (2016-12-25)
TAK1 (transforming growth factor-β-activated kinase 1) is an essential intracellular mediator of cytokine and growth factor signaling and a potential therapeutic target for the treatment of immune diseases and cancer. Herein we report development of a series of 2,4-disubstituted pyrimidine
Suman Rao et al.
Cell chemical biology, 26(6), 818-829 (2019-04-16)
Covalent kinase inhibitors, which typically target cysteine residues, represent an important class of clinically relevant compounds. Approximately 215 kinases are known to have potentially targetable cysteines distributed across 18 spatially distinct locations proximal to the ATP-binding pocket. However, only 40

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service