Skip to Content
Merck
All Photos(1)

Key Documents

SML1132

Sigma-Aldrich

Pep2-8 trifluoroacetate salt

≥95% (HPLC), contains 58μL of solution at 10mM in DMSO

Synonym(s):

Ac-TVFTSWEEYLDWV-NH2 trifluoroacetate salt, Ac-TVFTSWEEYLDWV-amide trifluoroacetate salt, Ac-Thr-Val-Phe-Thr-Ser-Trp-Glu-Glu-Tyr-Leu-Asp-Trp-Val-NH2 trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$243.00

$243.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$243.00

About This Item

Empirical Formula (Hill Notation):
C83H110N16O24 · xC2HF3O2
Molecular Weight:
1715.85 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

$243.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

solution

color

colorless to pale yellow

shipped in

wet ice

storage temp.

−20°C

Amino Acid Sequence

Ac-Thr-Val-Phe-Thr-Ser-Trp-Glu-Glu-Tyr-Leu-Asp-Trp-Val-NH2 trifluoroacetate salt

Biochem/physiol Actions

Pep2-8 is a potent PCSK9 (proprotein convertase subtilisin/kexin type 9) inhibitor that selectively binds to PCSK9 and interferes with LDL receptor binding to PCSK9. Pep2-8 restores LDL receptor function and LDL uptake of PCSK9-treated HepG2 cells.
Pep2-8 is a potent PCSK9 (proprotein convertase subtilisin/kexin type 9) inhibitor.
Pep2-8, a good mimic of epidermal growth factor precursor homology domain A (EGF-A)[1], serves as a competitive inhibitor of low-density lipoprotein (LDL) receptor binding.[2] It can be used as an anchor peptide in phage-display experiments to bind an extension peptide library to the groove site.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yingnan Zhang et al.
The Journal of biological chemistry, 289(2), 942-955 (2013-11-15)
PCSK9 (proprotein convertase subtilisin/kexin type 9) is a negative regulator of the hepatic LDL receptor, and clinical studies with PCSK9-inhibiting antibodies have demonstrated strong LDL-c-lowering effects. Here we screened phage-displayed peptide libraries and identified the 13-amino acid linear peptide Pep2-8
Yingnan Zhang et al.
Nature structural & molecular biology, 24(10), 848-856 (2017-08-22)
Proprotein convertase subtilisin/kexin type 9 (PCSK9) regulates plasma LDL cholesterol (LDL-c) levels by promoting the degradation of liver LDL receptors (LDLRs). Antibodies that inhibit PCSK9 binding to the EGF(A) domain of the LDLR are effective in lowering LDL-c. However, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service