Skip to Content
Merck
All Photos(1)

Documents

PHL80431

Dihydromyricetin

phyproof® Reference Substance

Synonym(s):

(2R,3R)-3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-2,3-dihydrochromen-4-one, 3,3′,4′,5,5′,7-Hexahydroxyflavanone, Ampelopsin, Ampeloptin, DHM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O8
CAS Number:
Molecular Weight:
320.25
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

SMILES string

O=C1C2=C(O)C=C(O)C=C2O[C@H](C3=CC(O)=C(C(O)=C3)O)[C@H]1O

InChI

1S/C15H12O8/c16-6-3-7(17)11-10(4-6)23-15(14(22)13(11)21)5-1-8(18)12(20)9(19)2-5/h1-4,14-20,22H/t14-,15+/m0/s1

InChI key

KJXSIXMJHKAJOD-LSDHHAIUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application


  • Hovenia dulcis: a Chinese medicine that plays an essential role in alcohol-associated liver disease. This review discusses the role of Hovenia dulcis, from which dihydromyricetin is derived, in treating alcohol-associated liver conditions, highlighting its mechanisms and therapeutic potentials (He YX, Liu MN, Wang YY, et al. 2024).

  • Dihydromyricetin ameliorates hepatic steatosis and insulin resistance via AMPK/PGC-1α and PPARα-mediated autophagy pathway. This study explores how dihydromyricetin influences liver health, particularly in hepatic steatosis and insulin resistance, offering insights into its mechanisms through autophagy pathways (Yang Y, Qiu W, Xiao J, et al. 2024).

  • Identification of dihydromyricetin as a natural DNA methylation inhibitor with rejuvenating activity in human skin. Research identifies dihydromyricetin′s potential anti-aging effects on human skin by modulating DNA methylation, which could contribute to its broader use in dermatological products (Falckenhayn C, Bienkowska A, Söhle J, et al. 2023).

  • Dihydromyricetin reverses capecitabine-induced peripheral myelin dysfunction through modulation of oxidative stress. This article provides evidence of dihydromyricetin′s protective effects against peripheral myelin damage due to oxidative stress, relevant in the treatment of certain neuropathies (Fang J, Lou S, Zhou X, et al. 2024).

  • The Molecular Mechanism Underlying the Therapeutic Effect of Dihydromyricetin on Type 2 Diabetes Mellitus Based on Network Pharmacology, Molecular Docking, and Transcriptomics. This comprehensive study details the molecular interactions and pathways through which dihydromyricetin could affect type 2 diabetes, providing a foundation for its application in metabolic disorder treatments (Wen X, Lv C, Zhou R, et al. 2024).

Biochem/physiol Actions

Dihydromyricetin (Ampelopsin) is a flavanonol with antioxidant and anti-cancer activity, found to have anti-alcohol intoxication effects. Its anti-alcohol effects appear to be by its actions as a positive modulator of GABA-A receptors at the benzodiazepine site.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service