Skip to Content
Merck
All Photos(1)

Key Documents

SML1010

Sigma-Aldrich

L-742001 hydrochloride

≥98% (HPLC)

Synonym(s):

(Z)-4-[1-Benzyl-4-[(4-chlorophenyl)methyl]piperidin-4-yl]-2-hydroxy-4-oxobut-2-enoic acid hydrochloride, 4-[4-[(4-Chlorophenyl)methyl]-1-(phenylmethyl)-4-piperidinyl]-2-hydroxy-4-oxo-2-butenoic acid hydrochloride, L-742,001 hydrochloride, L742001 hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$187.00
25 MG
$746.00

$187.00


Estimated to ship onApril 01, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$187.00
25 MG
$746.00

About This Item

Empirical Formula (Hill Notation):
C23H24ClNO4 · HCl
CAS Number:
Molecular Weight:
450.35
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$187.00


Estimated to ship onApril 01, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O/C(C(O)=O)=C\C(C(CC1)(CC2=CC=C(Cl)C=C2)CCN1CC3=CC=CC=C3)=O.Cl

InChI

1S/C23H24ClNO4.ClH/c24-19-8-6-17(7-9-19)15-23(21(27)14-20(26)22(28)29)10-12-25(13-11-23)16-18-4-2-1-3-5-18;/h1-9,14,26H,10-13,15-16H2,(H,28,29);1H/b20-14-;

InChI key

LMLNSXCRZNNZBT-VSOKSMTPSA-N

Biochem/physiol Actions

L-742,001 is an inhibitor of the influenza virus RNA polymerase PA subunit.
L-742,001 is an inhibitor of the influenza virus RNA polymerase PA subunit, implicated in several roles, including endonuclease and protease activities as well as viral RNA/complementary RNA promoter binding. L-742,001 inhibits the influenza endonuclease-dependent polymerase activity with an IC50 of 430 nM.
L-742001 is useful in the treatment of influenza viral infection.[1]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Binding affinity of the L-742,001 inhibitor to the endonuclease domain of A/H1N1/PA influenza virus variants: Molecular simulation approaches.
Nguyen H, et al.
Chemical Physics, 500, 26-36 (2018)
Wenjie Wang et al.
Cell reports, 30(1), 153-163 (2020-01-09)
Severe fever with thrombocytopenia syndrome virus (SFTSV) is a tick-borne virus with 12%-30% case mortality rates and is related to the Heartland virus (HRTV) identified in the United States. Together, SFTSV and HRTV are emerging segmented, negative-sense RNA viral (sNSV) pathogens with

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service