Skip to Content
Merck
All Photos(1)

Documents

P0040

Sigma-Aldrich

PP1

≥98% (HPLC)

Synonym(s):

4-Amino-5-(methylphenyl)-7-(t-butyl)pyrazolo-(3,4-d)pyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19N5
CAS Number:
Molecular Weight:
281.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >20 mg/mL

storage temp.

room temp

SMILES string

Cc1ccc(cc1)-c2nn(c3ncnc(N)c23)C(C)(C)C

InChI

1S/C16H19N5/c1-10-5-7-11(8-6-10)13-12-14(17)18-9-19-15(12)21(20-13)16(2,3)4/h5-9H,1-4H3,(H2,17,18,19)

InChI key

ZVPDNRVYHLRXLX-UHFFFAOYSA-N

Application

PP1 has been used as:
  • an inhibitor of sarcoma (Src) family kinases (SFK) like hematopoietic cell kinase (hck) and fyn
  • a selective Src tyrosine kinase inhibitor in hippocampal neuronal cultures to test its effect on neurite growth
  • a Src-kinase blocker to test its effect on brimonidine (BMD)-induced phosphorylation in extracellular signal-activated kinases(ERK1/2)

Biochem/physiol Actions

PP1 is a pyrazolopyrimidine compound that acts as a competitive inhibitor of adenosine triphosphate (ATP) binding. It also inhibits protein tyrosine kinase (PTK6) and may be useful in the therapeutic management of PTK6 positive based breast cancer malignancy.
PP1 is a potent and selective Src family protein tyrosine kinase inhibitor.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Shunyu Gao et al.
General and comparative endocrinology, 240, 46-60 (2016-10-23)
Neuropeptide Y (NPY) receptors and its ligands, NPY, peptide YY (PYY) and pancreatic polypeptide (PP), are suggested to regulate many physiological processes including food intake in birds. However, our knowledge regarding this avian NPY system remains rather limited. Here, we
Katyayni Vinnakota et al.
Journal of cellular physiology, 232(12), 3468-3480 (2017-01-18)
The inflammatory milieu plays an important role in colon cancer development and progression. Previously, we have shown that tumor-associated macrophages (TAMs), an important component of the tumor microenvironment, are enriched in tumors compared with normal tissue and confer a poorer
Qian Wang et al.
Environmental science and pollution research international, 26(21), 21774-21783 (2019-05-28)
Due to variable amino acid residues at positions 2 and 4, microcystins (MCs) had diversified variants with different toxicities. To evaluate the discrepant toxicity, the inhibition effects of five typical MC variants (with the changed amino acid residues at position
Ames C Register et al.
Methods in molecular biology (Clifton, N.J.), 1636, 79-89 (2017-07-22)
The Src family kinases (SFKs) are an important family of tyrosine kinases that are allosterically regulated by their SH2 and SH3 domains. Engagement of SFK SH2 and SH3 domains with their intramolecular ligands leads to reduced kinase activity by stabilizing
Jie Sun et al.
Nature communications, 8(1), 477-477 (2017-09-09)
Protein-based biosensors or activators have been engineered to visualize molecular signals or manipulate cellular functions. Here we integrate these two functionalities into one protein molecule, an integrated sensing and activating protein (iSNAP). A prototype that can detect tyrosine phosphorylation and

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service