Skip to Content
Merck
All Photos(1)

Key Documents

SML0545

Sigma-Aldrich

CID 1067700

≥98% (HPLC)

Synonym(s):

2-(Benzoylcarbamothioylamino)-5,5-dimethyl-4,7-dihydrothieno[2,3-c]pyran-3-carboxylic acid, 2-[[(Benzoylamino)thioxomethyl]amino]-4,7-dihydro-5,5-dimethyl-5H-thieno[2,3-c]pyran-3-carboxylic acid, CID-1067700, CID1067700, SID57578339

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$155.00
25 MG
$623.00

$155.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$155.00
25 MG
$623.00

About This Item

Empirical Formula (Hill Notation):
C18H18N2O4S2
CAS Number:
Molecular Weight:
390.48
UNSPSC Code:
12352200
NACRES:
NA.77

$155.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

[s]1c2c(c(c1NC(=S)NC(=O)c3ccccc3)C(=O)O)CC(OC2)(C)C

InChI

1S/C18H18N2O4S2/c1-18(2)8-11-12(9-24-18)26-15(13(11)16(22)23)20-17(25)19-14(21)10-6-4-3-5-7-10/h3-7H,8-9H2,1-2H3,(H,22,23)(H2,19,20,21,25)

InChI key

ATSWBWHRHAQVFM-UHFFFAOYSA-N

Biochem/physiol Actions

CID 1067700 antagonizes ras-related protein Rab-7 and reduces class switch DNA recombination (CSR) in B cells and survival of plasma cells.[1]
CID 1067700 is a potent and competitive inhibitor of Ras-related GTPases that potently binds to Rab7 nucleotide binding site.
CID 1067700 is a potent and competitive inhibitor of Ras-related GTPases.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Remedy for Lupus through Restraining Rab7
How H I V.
Journal of Immunology, 197, 3749-3750 (2016)
Shuai Li et al.
Experimental cell research, 370(1), 87-97 (2018-06-17)
Autophagy dysfunction has been observed in intervertebral disc degeneration (IVDD) cells, a main contributing factor to cell death, but the precise role of autophagy during IVDD is still controversial. This study aimed to investigate the role of autophagy involved in
Lin Bai et al.
Cell death & disease, 12(4), 392-392 (2021-04-14)
Amyotrophic lateral sclerosis (ALS) is a progressive neurodegenerative disease caused by motoneuron loss, for which there is currently no effective treatment. Statins, as inhibitors of 3-hydroxy-3-methylglutaryl-CoA (HMG-CoA) reductase, are used as drugs for treatment for a variety of disease such
Xueyu Wang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(11), 14473-14489 (2020-09-07)
O-GlcNAcylation is a form of posttranslational modification, and serves various functions, including modulation of location, stability, and activity for the modified proteins. O-linked-N-acetylglucosamine (O-GlcNAc) transferase (OGT) is an essential cellular enzyme that posttranslationally modifies the cellular proteins with O-GlcNAc moiety.
Yong Lin et al.
Autophagy, 16(3), 548-561 (2019-06-18)
Glucosamine (GlcN), a dietary supplement widely utilized to promote joint health and effective in the treatment of osteoarthritis, is an effective macroautophagy/autophagy activator in vitro and in vivo. Previous studies have shown that autophagy is required for hepatitis B virus

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Questions

  1. After resuspending 5 mg CID 1067700 in DMSO, should it still be stored at 4°C or should it be stored at -20°C?

    1 answer
    1. The stability of this material in solution at 4°C has not been determined. Following reconstitution, aliquot and freeze at -20°C for up to 3 months.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service