Skip to Content
Merck
All Photos(1)

Key Documents

SML3126

Sigma-Aldrich

BI-2852

≥98% (HPLC)

Synonym(s):

(3S)-2,3-Dihydro-5-hydroxy-3-[2-[[[[1-[(1-methyl-1H-imidazol-4-yl)methyl]-1H-indol-6-yl]methyl]amino]methyl]-1H-indol-3-yl]-1H-isoindol-1-one, (S)-5-Hydroxy-3-(2-(((1-((1-methyl-1H-imidazol-4-yl)methyl)-1H-indol-6-yl)methylamino)methyl)-1H-indol-3-yl)isoindolin-1-one, BI 2852, BI2852

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$425.00
25 MG
$1,690.00

$425.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$425.00
25 MG
$1,690.00

About This Item

Empirical Formula (Hill Notation):
C31H28N6O2
CAS Number:
Molecular Weight:
516.59
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$425.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1C2=C([C@H](N1)C3=C(NC4=C3C=CC=C4)CNCC5=CC6=C(C=C5)C=CN6CC7=CN(C=N7)C)C=C(C=C2)O

InChI key

JYEQLXOWWLNVDX-PMERELPUSA-N

Biochem/physiol Actions

BI-2852 is RAS inhibitor that binds RAS switch I and II (SI/II) pocket in both the active and inactive forms (GCP-/GDP-bound KD in μM = 0.74/2.0 (KRAS-G12D), 7.5/1.1 (KRAS), 0.57/2.5 (HRAS), 1.3/8.3 (NRAS)). BI-2852 inhibits RAS-mediated signaling and proliferation of KRAS-G12C-bearing NCI-H358 lung cancer cells (EGF-induced pERK IC50 = 5.8 μM; GI50 = 6.7 μM in soft agar with low serum) by blocking KRAS interactions with GEF SOS1 (GDP-bound G12C/G12D IC50 = 450/260 nM, GTP-bound Wt/G12C/G12D IC50 = 490/360/490 nM) and effectors CRAF (GTP-bound Wt/G12C/G12D IC50 = 1740/80/770 nM) and PI3Kα (GTP-bound Wt/G12C/G12D IC50 = 250/100/500 nM).
RAS switch I and II (SI/II) pocket-targeting inhibitor against RAS-mediated signaling and proliferation by blocking KRAS interactions with GEF and effectors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dirk Kessler et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(32), 15823-15829 (2019-07-25)
The 3 human RAS genes, KRAS, NRAS, and HRAS, encode 4 different RAS proteins which belong to the protein family of small GTPases that function as binary molecular switches involved in cell signaling. Activating mutations in RAS are among the
The small molecule BI-2852 induces a nonfunctional dimer of KRAS.
Timothy H Tran et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(7), 3363-3364 (2020-02-13)
Reply to Tran et al.: Dimeric KRAS protein-protein interaction stabilizers.
Dirk Kessler et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(7), 3365-3367 (2020-02-13)
Rachel Cooley et al.
Wellcome open research, 5, 20-20 (2020-06-27)
Targeting the interaction of proteins with weak binding affinities or low solubility represents a particular challenge for drug screening. The NanoLuc â ® Binary Technology (NanoBiT â ®) was originally developed to detect protein-protein interactions in live mammalian cells. Here we report

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service