S5643
Sulfanilic acid
ACS reagent, 98.0-102.0%
Synonym(s):
4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid
About This Item
Recommended Products
grade
ACS reagent
Assay
98.0-102.0%
impurities
≤0.02% Insoluble in sodium carbonate solution
ign. residue
≤0.01%
mp
>300 °C (lit.)
anion traces
chloride (Cl-): ≤0.002%
nitrite (NO2-): ≤0.5 ppm
sulfate (SO42-): ≤0.01%
SMILES string
Nc1ccc(cc1)S(O)(=O)=O
InChI
1S/C6H7NO3S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H,8,9,10)
InChI key
HVBSAKJJOYLTQU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service