Skip to Content
Merck
All Photos(1)

Key Documents

CRM47791

Supelco

Linoleic Acid Methyl Ester Mix

cis/trans, certified reference material, 10 mg/mL in dichloromethane

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$204.00

$204.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 24, 2025Details


Request a Bulk Order

Select a Size

Change View
1 ML
$204.00

About This Item

EC Number:
UNSPSC Code:
85151701
NACRES:
NA.24

$204.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 24, 2025Details


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

liquid

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

10 mg/mL in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

single component solution

functional group

carboxylic acid
ester

storage temp.

-10 to -25°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

The CRM has been used as follows:

  • To study the retention of two FAME mixtures on three types of available commercial capillary columns having different polarities for developing a gas chromatography-mass spectrometry (GC-MS) based method to separate geometrical isomers of polyunsaturated fatty acids (PUFAs)
  • Development and validation of a GC-MS method to simultaneously determine C18 fatty acids in 20 raw milk and 30 commercial milk samples
  • Identification of fatty acid methyl ester peaks in the GC-MS analysis of C18 fatty acids in milk samples to evaluate the hydrogenation of α-Linolenic acid in the rumen of dairy cows
  • Determination of fatty acids following their methyl esterification in goat milk samples by gas chromatography coupled to a flame ionization detector (GC-FID) in the study to investigate the effect of production systems on the fatty acid composition of goat milk
  • GC-FID analysis of typical Indian ready-made snacks to determine their fatty acid composition based on Association of Official Analytical Chemists (AOAC) Official Method 996.06
  • To study the fatty acid profile of 69 different cold-pressed vegetable oil samples by gas chromatography coupled to a flame ionization detector

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    cis-9,cis-12-Octadecadienoic acid methyl ester 10 % (w/w)
    cis-9,trans-12-Octadecadienoic acid methyl ester 20 % (w/w)
    trans-9,cis-12-Octadecadienoic acid methyl ester 20 % (w/w)
    trans-9,12-Octadecadienoic acid methyl ester 50 % (w/w)

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Enriquez-Hidalgo et al.
Frontiers in veterinary science, 7, 576944-576944 (2020-12-18)
The winter diet of dairy cows in Mediterranean climate regions is usually a total mixed ration with a base of conserved summer crops such as corn silage and alfalfa hay. However, there is increased labor and financial cost related to
Arkadiusz Szterk et al.
Foods (Basel, Switzerland), 11(8) (2022-04-24)
The study aimed to examine samples of different market original sheep cow and goat cheeses, in respect of the content and profile of FA with special emphasis on health-promoting FA. The content of fatty acids in the examined cheeses was

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How is the standard prepared for injection into a gas chromatograph ?

    1 answer
    1. This product is provided as a ready-to-use solution, at a nominal concentration of 10 mg/mL, so no further preparation is necessary. The typical injection volume for the GC is 1 microliter.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service