Skip to Content
Merck
All Photos(3)

Key Documents

251917

Sigma-Aldrich

Sulfanilic acid

ACS reagent, 99%

Synonym(s):

4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$42.70
100 G
$135.00
500 G
$427.00

$42.70


Estimated to ship onMarch 20, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$42.70
100 G
$135.00
500 G
$427.00

About This Item

Linear Formula:
4-(H2N)C6H4SO3H
CAS Number:
Molecular Weight:
173.19
Beilstein:
908765
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

$42.70


Estimated to ship onMarch 20, 2025


Request a Bulk Order

grade

ACS reagent

Quality Level

Assay

98.0-102.0% (ACS specification)
99%

form

powder

ign. residue

≤0.01%

mp

>300 °C (lit.)

anion traces

chloride (Cl-): ≤0.002%
nitrite (NO2-): ≤0.5 ppm
sulfate (SO42-): ≤0.01%

functional group

sulfonic acid

SMILES string

Nc1ccc(cc1)S(O)(=O)=O

InChI

1S/C6H7NO3S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H,8,9,10)

InChI key

HVBSAKJJOYLTQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The diazonium salt of sulfanilic acid may be used in the preparation of azo dyes such as o-anisaldehyde[1], orange I[2] and orange II.[2] It may also be used in the preparation of 2,6-dibromoaniline via bromination followed by desulfonation.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1,2-AMINONAPHTHOL HYDROCHLORIDE
Conant JB, et al
Organic Syntheses, 17, 9-9 (1937)
2,6-DICHLOROANILINE AND 2,6-DIBROMOANILINE
Seikel MK
Organic Syntheses, 24, 47-47 (1944)
o-ANISALDEHYDE
Sisti AJ
Organic Syntheses, 44, 4-4 (1964)
Adsorption of essential micronutrients by carbon nanotubes and the implications for nanotoxicity testing.
Lin Guo et al.
Small (Weinheim an der Bergstrasse, Germany), 4(6), 721-727 (2008-05-28)
Kinetic, mechanistic and spectral studies for the oxidation of sulfanilic acid by alkaline hexacyanoferrate (III).
Farokhi SA and Nandibewoor ST.
Tetrahedron, 59(38), 7595-7602 (2003)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service