Skip to Content
Merck
All Photos(1)

Key Documents

S2771

Scalpel blades

No. 11

Synonym(s):

Surgical Scalpel Blades

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$270.00

$270.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
100 EA
$270.00

About This Item

UNSPSC Code:
41100000
NACRES:
NB.35

$270.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

steel (carbon)

sterility

non-sterile

Looking for similar products? Visit Product Comparison Guide

Application

Scalpel blades have been used:
  • for the generation of organoids
  • to perform rodent nucleus accumbens (NAc) manual microdissection
  • to trim embryonic brain sample

Packaging

individually wrapped.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service