Skip to Content
Merck
All Photos(1)

Documents

Z265977

Dissecting scissors

L 4 1/2 in., Sharp, straight

Synonym(s):

Surgical scissors, cutting tool

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122408
NACRES:
NB.35

packaging

pack of 1 ea

L

4 1/2 in.

Looking for similar products? Visit Product Comparison Guide

General description

Stainless steel dissecting instruments

Application

Dissecting scissors have been used to cut the stem of Arabidopsis during the separation of the male gametophyte.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service