Skip to Content
Merck
All Photos(6)

Key Documents

S2626

Sigma-Aldrich

Spermidine

Spermidine
1 of 1 reviewers received a sample product or took part in a promotion

≥99% (GC)

Synonym(s):

1,8-Diamino-4-azaoctane, N-(3-Aminopropyl)-1,4-diaminobutane

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$131.00
5 G
$235.00
25 G
$937.00

$131.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$131.00
5 G
$235.00
25 G
$937.00

About This Item

Linear Formula:
NH2(CH2)3NH(CH2)4NH2
CAS Number:
Molecular Weight:
145.25
Beilstein:
1698591
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

$131.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

synthetic

Assay

≥99% (GC)

form

solid or liquid (clear)

impurities

DNase, none detected
RNase, none detected
protease, none detected

refractive index

n20/D 1.479 (lit.)

mp

22-25 °C

solubility

water: 50 mg/mL, clear to very slightly hazy, colorless to very faintly yellow

density

0.925 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

NCCCCNCCCN

InChI

1S/C7H19N3/c8-4-1-2-6-10-7-3-5-9/h10H,1-9H2

InChI key

ATHGHQPFGPMSJY-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Spermidine, a natural biomolecule, is known for its wide-ranging health benefits, including anti-inflammatory effects, antioxidant properties, and improvements in respiratory function. It carries a positive charge at physiological pH, allowing it to interact with oppositely charged DNA and RNA. Dietary intake of spermidine is associated with a reduced risk of neurodegeneration, metabolic diseases, heart ailments, and cancer. Spermidine-induced autophagy has been shown to slow the rate of cognitive decline by clearing amyloid-beta plaques in the brain. Spermidine supplementation improves mitochondrial metabolism and translational activity. Moreover, spermidine exhibits cell-autonomous effects on cancer cells and influences their communication with immune effectors, leading to the acceleration of tumor-associated antigen identification and cancer cell death. Spermidine-mediated autophagy inhibits the activation of apoptosis in cancer cells and is linked to tissue development and cell differentiation. This natural compound is additionally recognized for its neuroprotective, and anti-aging properties and is naturally found in semen, as well as in various plant sources.[1]
The enzyme nitric oxide synthase (nNOS) is involved in the production of nitric oxide.[2]

Application

Spermidine has been used:
  • in the preparation of 5x transcription buffer[3]
  • in the treatment of double-stranded RNA along with other reagents to increase the uptake of dsRNA by nematodes[4]
  • as a standard in the analytical study of rat brain polyamines (185)

Biochem/physiol Actions

Inhibits neuronal nitric oxide synthase (nNOS). Binds and precipitates DNA; Stimulates T4 polynucleotide kinase activity.
Inhibits neuronal nitric oxide synthase (nNOS). Binds and precipitates DNA; may be used for purification of DNA binding proteins. Stimulates T4 polynucleotide kinase activity.

Caution

Solutions should be sterile-filtered, not autoclaved, if sterile solution is necessary. Spermidine deaminates with time; solutions should be stored frozen. Prepare new solutions frequently.

Other Notes

Store under inert gas. Air sensitive; strongly hygroscopic.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spermidine as a promising anticancer agent: Recent advances and newer insights on its molecular mechanisms
Parteek P, et al.
Frontiers in Chemistry, 11, 1164477-1164477 (2023)
In vitro Reconstitution Assay of miRNA Biogenesis by Arabidopsis DCL1.
Wang T
Bio-protocol, 5(8), e1454-e1454 (2015)
Evidence for the contribution of NOS1 gene polymorphism (rs3782206) to prefrontal function in schizophrenia patients and healthy controls.
Zhang Z
Neuropsychopharmacology, 40(6), 1383-1394 (2015)
Michi Miura et al.
Chromosome research : an international journal on the molecular, supramolecular and evolutionary aspects of chromosome biology, 28(3-4), 381-393 (2020-10-19)
CUT&RUN is a powerful tool to study protein-DNA interactions in vivo. DNA fragments cleaved by the targeted micrococcal nuclease identify the footprints of DNA-binding proteins on the chromatin. We performed CUT&RUN on human lung carcinoma cell line A549 maintained in
Quantitative Detection of Double-Stranded RNA-Mediated Gene Silencing of Parasitism Genes in Heterodera glycines.
Sukno SA
Journal of Nematology, 39(2), 145-152 (2007)

Questions

1–5 of 5 Questions  
  1. We have purchased S2626 spermidine 5 g. It does not say anywhere what molar concentration or if it is actually dissolved in water. Could you please inform me about that. I received 5 g i aprox 4 ml, and as I can read, the solubility is 50 mg /ml. ???

    1 answer
    1. This product is supplied as a neat or pure liquid with a density of 0.925 g/mL and a molecular weight of 145.25 g/mol. The minimum purity is 99%, thus the molarity will vary slightly from lot to lot. The molarity can range from 6.3 M ( 99%) to 6.4 M (100%). The lot-specific purity is reported on the Certificate of Analysis.

      Helpful?

  2. Is the Molarity for all Spermidin batches the same ? Does the 6,4 M count also for xx-1G and xx-25G? 

    1 answer
    1. The molarity of this compound is the same for all pack sizes, but may vary slightly from lot to lot. The reported 6.4 M is assuming a 100% purity. The minimum purity specification is 99%.

      Helpful?

  3. What is the molarity of Spermidine S2626-5G solution and what is the longterm storage condition?

    1 answer
    1. This product is a neat or pure liquid. Based on a density of 0.925 g/mL and a molecular weight of 145.25, this material has an effective molarity of 6.4 M. It is recommended to store this solution as single-use aliquots at -20 ºC for no more than one month.

      Helpful?

  4. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  5. Is this a spermidine solution or a neat liquid, and what is the concentration?

    1 answer
    1. Based on a density of 0.925 g/mL and a molecular weight of 145.25, this neat liquid has an effective molarity of 6.4 M.

      Helpful?

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. Buhler, Kansas
    • Review 1
    • Vote 1
    5 out of 5 stars.

    High Quality Product and Service

    This was a high quality product that arrived fast and with very good packaging. The product made our project a success and we were all very happy with the results!

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service