Skip to Content
Merck
All Photos(1)

Key Documents

S0381

Sigma-Aldrich

Spermidine phosphate salt hexahydrate

Synonym(s):

Dispermidine triphosphate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$125.00

$125.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$125.00

About This Item

Linear Formula:
[NH2(CH2)3NH(CH2)4NH2]2 · 3H3PO4 · 6H2O
CAS Number:
Molecular Weight:
346.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$125.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

storage temp.

−20°C

SMILES string

O.O.O.O.O.O.OP(O)(O)=O.OP(O)(O)=O.OP(O)(O)=O.NCCCCNCCCN.NCCCCNCCCN

InChI

1S/2C7H19N3.3H3O4P.6H2O/c2*8-4-1-2-6-10-7-3-5-9;3*1-5(2,3)4;;;;;;/h2*10H,1-9H2;3*(H3,1,2,3,4);6*1H2

InChI key

UYNKOICVVIGVMD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibits neuronal nitric oxide synthase (nNOS). Binds and precipitates DNA, may be used for purification of DNA binding proteins. Stimulates T4 polynucleotide kinase activity.

Caution

Solutions should be sterile-filtered, not autoclaved, if sterile solution is necessary. Spermidine deaminates with time; solutions should be stored frozen. Prepare new solutions frequently.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service