Skip to Content
Merck
All Photos(1)

Key Documents

709891

Sigma-Aldrich

Spermidine-(butyl-d8) trihydrochloride

98 atom % D, 95% (CP)

Synonym(s):

Deuterated spermidine trihydrochloride, N-·(3-·Aminopropyl)·-1,4-·butane-d8-diamine trihydrochloride, N-(3-Aminopropyl)-1,4-butane-d8-diamine trihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$1,640.00
10 MG
$2,940.00

$1,640.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request more information

Select a Size

Change View
5 MG
$1,640.00
10 MG
$2,940.00

About This Item

Linear Formula:
NH2(CH2)3NH(CD2)4NH2⋅3HCl
CAS Number:
Molecular Weight:
262.68
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.24

$1,640.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request more information

isotopic purity

98 atom % D

Assay

95% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mp

257-259 °C (lit.)

mass shift

M+8

storage temp.

−20°C
−20°C

SMILES string

Cl.Cl.Cl.[2H]C([2H])(N)C([2H])([2H])C([2H])([2H])C([2H])([2H])NCCCN

InChI

1S/C7H19N3.3ClH/c8-4-1-2-6-10-7-3-5-9;;;/h10H,1-9H2;3*1H/i1D2,2D2,4D2,6D2;;;

InChI key

LCNBIHVSOPXFMR-BQVYHINQSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service