Skip to Content
Merck
All Photos(1)

Key Documents

S0763

Sigma-Aldrich

N-Succinyl-Ala-Phe-Lys 7-amido-4-methylcoumarin acetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$217.00
25 MG
$818.00

$217.00


Estimated to ship onApril 20, 2025



Select a Size

Change View
5 MG
$217.00
25 MG
$818.00

About This Item

Empirical Formula (Hill Notation):
C32H39N5O8 · C2H4O2
CAS Number:
Molecular Weight:
681.73
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

$217.00


Estimated to ship onApril 20, 2025


Assay

≥95% (TLC)

Quality Level

form

powder

solubility

1 M NH4OH: 50 mg/mL, clear to slightly hazy, colorless to light yellow

storage temp.

−20°C

SMILES string

CC(O)=O.C[C@H](NC(=O)CCC(O)=O)C(=O)N[C@@H](Cc1ccccc1)C(=O)N[C@@H](CCCCN)C(=O)Nc2ccc3C(C)=CC(=O)Oc3c2

InChI

1S/C32H39N5O8.C2H4O2/c1-19-16-29(41)45-26-18-22(11-12-23(19)26)35-31(43)24(10-6-7-15-33)36-32(44)25(17-21-8-4-3-5-9-21)37-30(42)20(2)34-27(38)13-14-28(39)40;1-2(3)4/h3-5,8-9,11-12,16,18,20,24-25H,6-7,10,13-15,17,33H2,1-2H3,(H,34,38)(H,35,43)(H,36,44)(H,37,42)(H,39,40);1H3,(H,3,4)/t20-,24-,25-;/m0./s1

InChI key

YSFJDCMZVSMICC-FMQWQMCSSA-N

Substrates

Fluorogenic substrate for plasmin

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pierzchala, P.A., et al.
The Biochemical Journal, 183, 155-155 (1979)
Ritika Puri et al.
Foods (Basel, Switzerland), 9(11) (2020-11-27)
The effects of processing temperature on filtration performance and characteristics of retentates and permeates produced during ultrafiltration (UF) of skim milk at 5, 20, and 50 °C were investigated. The results indicate that despite higher flux at 50 °C, UF

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service