Skip to Content
Merck
All Photos(1)

Documents

B4636

Sigma-Aldrich

Boc-Leu-Ser-Thr-Arg-7-amido-4-methylcoumarin

protein C substrate, ≥98% (HPLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H52N8O10
CAS Number:
Molecular Weight:
732.82
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

product name

Boc-Leu-Ser-Thr-Arg-7-amido-4-methylcoumarin, protein C substrate

Assay

≥98% (HPLC)

form

powder

solubility

ethanol: 2 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)OC(C)(C)C)C(=O)NC(CO)C(=O)NC(C(C)O)C(=O)NC(CCCNC(N)=N)C(=O)Nc1ccc2C(C)=CC(=O)Oc2c1

InChI

1S/C34H52N8O10/c1-17(2)13-23(41-33(50)52-34(5,6)7)29(47)40-24(16-43)30(48)42-27(19(4)44)31(49)39-22(9-8-12-37-32(35)36)28(46)38-20-10-11-21-18(3)14-26(45)51-25(21)15-20/h10-11,14-15,17,19,22-24,27,43-44H,8-9,12-13,16H2,1-7H3,(H,38,46)(H,39,49)(H,40,47)(H,41,50)(H,42,48)(H4,35,36,37)

InChI key

ZJNWKMRGENOMCV-UHFFFAOYSA-N

Related Categories

Substrates

A fluorogenic substrate for activated protein C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Sawada et al.
The Journal of experimental zoology, 254(1), 83-87 (1990-04-01)
Enzymatic properties of a protease involved in hatching of mouse embryos were examined. A trypsin-like protease, which most efficiently hydrolyzed t-butoxycarbonyl-Leu-Ser-Thr-Arg-4-methylcoumaryl-7-amide, was demonstrated in culture medium of mouse hatching embryos. The enzyme was strongly inhibited by diisopropyl fluorophosphate, phenylmethanesulfonyl fluoride
Brian T O'Neill et al.
The Journal of clinical investigation, 126(9), 3433-3446 (2016-08-16)
Diabetes strongly impacts protein metabolism, particularly in skeletal muscle. Insulin and IGF-1 enhance muscle protein synthesis through their receptors, but the relative roles of each in muscle proteostasis have not been fully elucidated. Using mice with muscle-specific deletion of the
Y Ohno et al.
Journal of biochemistry, 90(5), 1387-1395 (1981-11-01)
Protein C is a precursor of plasma serine proteinases, and its active form inactivates specifically blood coagulation Factor V and Factor VIII. Since a specific and sensitive synthetic substrate for the activated protein C was not known, we studied its
Ming Li et al.
Ecotoxicology and environmental safety, 135, 24-31 (2016-09-28)
Avermectins (AVMs) are used worldwide in agriculture and veterinary medicine. Residues of avermectin drugs, causing toxicological effects on non-target organisms, have raised great concern. The aim of this study was to investigate the effects of AVM on the expression levels
M A Nurseitova et al.
Journal of dairy science, 102(9), 7723-7733 (2019-07-01)
Milk fat adulteration is a common issue in Central Asia. To assess the current situation in the commercial milk market, 17 milk samples were checked for fatty acid (FA) and sterol profiles to detect potential adulteration using multivariate analysis. Analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service