Skip to Content
Merck
All Photos(1)

Key Documents

C8022

Sigma-Aldrich

Z-L-Arg 7-amido-4-methylcoumarin hydrochloride

Synonym(s):

Carbobenzoxy-L-arginine-7-amino-4-methylcoumarin hydrochloride, Nα-Cbz-L-arginine 7-amido-4-methylcoumarin hydrochloride, Z-L-Arginine-4-methyl-7-coumarinylamide hydrochloride, Z-Arg-MCA

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$565.00

$565.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025



Select a Size

Change View
50 MG
$565.00

About This Item

Empirical Formula (Hill Notation):
C24H27N5O5 · HCl
CAS Number:
Molecular Weight:
501.96
Beilstein:
4637954
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

$565.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Assay

≥98% (TLC)

form

powder

solubility

methanol: 20 mg/mL, clear, colorless to faintly yellow

fluorescence

λex 290 nm; λem 390 nm (buffer pH 7.1)
λex 290 nm; λem 437 nm (Reaction product)

storage temp.

−20°C

SMILES string

Cl[H].CC1=CC(=O)Oc2cc(NC(=O)[C@H](CCCNC(N)=N)NC(=O)OCc3ccccc3)ccc12

InChI

1S/C24H27N5O5.ClH/c1-15-12-21(30)34-20-13-17(9-10-18(15)20)28-22(31)19(8-5-11-27-23(25)26)29-24(32)33-14-16-6-3-2-4-7-16;/h2-4,6-7,9-10,12-13,19H,5,8,11,14H2,1H3,(H,28,31)(H,29,32)(H4,25,26,27);1H/t19-;/m0./s1

InChI key

JYWCRDUTYUBXOL-FYZYNONXSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service