Skip to Content
Merck
All Photos(1)

Documents

PZ0110

Sigma-Aldrich

SC-57461A

≥98% (HPLC)

Synonym(s):

N-Methyl-N-[3-[4-(phenylmethyl)phenoxy]propyl β-alanine hydrochloride, SC-57461 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H25NO3 · HCl
CAS Number:
Molecular Weight:
363.88
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >5 mg/mL

storage temp.

2-8°C

SMILES string

Cl.CN(CCCOc1ccc(Cc2ccccc2)cc1)CCC(O)=O

InChI

1S/C20H25NO3.ClH/c1-21(14-12-20(22)23)13-5-15-24-19-10-8-18(9-11-19)16-17-6-3-2-4-7-17;/h2-4,6-11H,5,12-16H2,1H3,(H,22,23);1H

InChI key

SBBJJLDNKNNWFJ-UHFFFAOYSA-N

Biochem/physiol Actions

SC-57461A is an inhibitor of leukotriene A4 hydrolase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to leukotriene receptors for your research needs.

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service