Skip to Content
Merck

Skip To

08084

Stearoyl-L-carnitine

analytical standard

Synonym(s):

(2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxooctadecyl)oxy]-1-propanaminium inner salt, L-Carnitine octadecanoyl ester, C18-Carnitine, Octadecanoyl-L-carnitine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Check Cart for Availability
$278.00

About This Item

Empirical Formula (Hill Notation):
C25H49NO4
CAS Number:
Molecular Weight:
427.66
UNSPSC Code:
41116107
NACRES:
NB.21
Beilstein/REAXYS Number:
7309670

$278.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥97.0% (HPLC)

optical activity

[α]/D -15.0±2.0°, c = 1 in methanol

shelf life

limited shelf life, expiry date on the label

impurities

≤10.0% water

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

C[N+](C)(C)C[C@H](OC(CCCCCCCCCCCCCCCCC)=O)CC([O-])=O

InChI

1S/C25H49NO4/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-25(29)30-23(21-24(27)28)22-26(2,3)4/h23H,5-22H2,1-4H3/t23-/m1/s1

InChI key

FNPHNLNTJNMAEE-HSZRJFAPSA-N

General description

Stearoyl-L-carnitine is a naturally occurring compound belonging to the acylcarnitine class of compounds as listed in the human metabolome database (HMBD). These compounds are reported to be used as biomarkers in the diagnosis of carnitine deficiency. It acts as a carrier of long chain acyl groups from activated fatty acids across the inner mitochondrial membrane into the mitochondrial matrix where they undergo β-oxidation to acetyl CoA to obtain usable energy via the citric acid cycle.

Application

Stearoyl-L-carnitine may be used as an analytical reference standard for the quantification of the analyte in biological fluids[1] using chromatography-based techniques.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
612290898406206
grade

analytical standard

grade

-

grade

analytical standard

grade

analytical standard

application(s)

clinical testing

application(s)

-

application(s)

clinical testing

application(s)

clinical testing

format

neat

format

-

format

neat

format

neat

assay

≥97.0% (HPLC)

assay

≥97.0% (TLC)

assay

≥97.0% (HPLC)

assay

≥97.0% (HPLC)

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

impurities

≤10.0% water

impurities

≤10% water

impurities

≤10.0% water

impurities

≤10% water


Still not finding the right product?

Explore all of our products under Stearoyl-L-carnitine


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters