Skip to Content
Merck
All Photos(1)

Key Documents

1368201

USP

Alpha Lipoic Acid

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(±)-α-Lipoic acid, (±)-1,2-Dithiolane-3-pentanoic acid, 6,8-Dithiooctanoic acid, DL-α-Lipoic acid, DL-6,8-Thioctic acid, Lip(S2)

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$491.00

$491.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$491.00

About This Item

Empirical Formula (Hill Notation):
C8H14O2S2
CAS Number:
Molecular Weight:
206.33
Beilstein:
81853
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$491.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

alpha lipoic acid

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)CCCCC1CCSS1

InChI

1S/C8H14O2S2/c9-8(10)4-2-1-3-7-5-6-11-12-7/h7H,1-6H2,(H,9,10)

InChI key

AGBQKNBQESQNJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Alpha Lipoic Acid USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

Antioxidant and coenzyme needed for the activity of enzyme complexes such as pyruvate dehydrogenase and glycine decarboxylase.
Antioxidant and coenzyme needed for the activity of enzyme complexes such as those of pyruvate dehydrogenase and glycine decarboxylase. Exogenous thioctic acid is reduced intracellularly by two or more enzymes. The reduced form then influences a number of cell processes by direct radical scavenging, recycling of other antioxidants, increasing glutathione synthesis, and modulating transcription factor activity particularly that of NF-κB. Decreases the phagocytosis of myelin by macrophages.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hee-jae Kim et al.
Neurochemical research, 39(8), 1579-1584 (2014-06-06)
Anti-convulsant effects of physical exercise and lipoic acid (LA), also referred to as thioctic acid with antioxidant activity, were investigated using chemical induced seizure model. We investigated the synergic effect of physical exercise and LA on kainic acid-induced seizure activity
Alexander Lossos et al.
Neurology, 84(7), 659-667 (2015-01-23)
To present the clinical, molecular, and cell biological findings in a family with an autosomal recessive form of hereditary spastic paraplegia characterized by a combination of spastic paraplegia, optic atrophy, and peripheral neuropathy (SPOAN). We used a combination of whole-genome
Alessandro Ghezzo et al.
PloS one, 9(11), e113111-e113111 (2014-11-25)
Down Syndrome (DS) is characterised by premature aging and an accelerated decline of cognitive functions in the vast majority of cases. As the life expectancy of DS persons is rapidly increasing, this decline is becoming a dramatic health problem. The
Yuzhe Tang et al.
Molecular cancer therapeutics, 13(6), 1526-1536 (2014-04-02)
Prostate cancers at the late stage of castration resistance are not responding well to most of current therapies available in clinic, reflecting a desperate need of novel treatment for this life-threatening disease. In this study, we evaluated the anticancer effect
Sander M Houten et al.
Human molecular genetics, 23(18), 5009-5016 (2014-05-23)
Dienoyl-CoA reductase (DECR) deficiency with hyperlysinemia is a rare disorder affecting the metabolism of polyunsaturated fatty acids and lysine. The molecular basis of this condition is currently unknown. We describe a new case with failure to thrive, developmental delay, lactic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service