Skip to Content
Merck
All Photos(1)

Key Documents

MAK319

Sigma-Aldrich

Isocitrate Assay Kit

sufficient for 100 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

application(s)

cosmetics
food and beverages

detection method

fluorometric

storage temp.

−20°C

General description

Isocitrate (Isocitric Acid) is a substrate in the citric acid (TCA) cycle. Isocitrate is formed by the isomerization of citrate catalyzed by the enzyme aconitase. Isocitrate is oxidized by isocitrate dehydrogenase producing α-ketoglutarate and generating NADPH. Isocitrate is commonly found in many fruits and vegetables and their processed products. Industrially, isocitrate is used as a marker to identify the quality and purity of fruit juices.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the quantitative determination of isocitrate and evaluation of drug effects on its metabolism in food, beverage and biological samples (e.g. cell lysate, tissue homogenate, serum, etc.).

Principle

This Isocitrate Assay Kit measures the NADPH generated from the oxidation of isocitrate. The NADPH reduces a probe into a highly fluorescent product. The fluorescence intensity of this product, measured at ex = 530 nm/ em = 585 nm, is proportional to the isocitrate concentration in the sample.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ya-Nan Xue et al.
Cancer medicine, 8(5), 2462-2473 (2019-04-12)
Metabolic reprogramming is a central hallmark of cancer. Therefore, targeting metabolism may provide an effective strategy for identifying promising drug targets for cancer treatment. In prostate cancer, cells undergo metabolic transformation from zinc-accumulating, citrate-producing cells to citrate-oxidizing malignant cells with

Questions

  1. What Isoform of Isocitrate is the Standard in the kit: D, L or DL?

    1 answer
    1. Isocitrate standard is DL-Isocitric acid.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service