Skip to Content
Merck
All Photos(1)

Key Documents

MAK443

Sigma-Aldrich

L-Lactate Assay Kit

Sufficient for 100 fluorometric tests

Synonym(s):

ʟ-Lactic Acid Assay Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$623.00

$623.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$623.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

$623.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

input

cell culture medium
plasma
serum
urine

detection method

fluorometric

relevant disease(s)

diabetes; pulmonary disorders; endocrinological disorders; cardiovascular diseases; cancer

storage temp.

−20°C

General description

Lactate is generated by lactate dehydrogenase (LDH) under hypoxic or anaerobic conditions. Monitoring lactate levels is, therefore, a good indicator of the balance between tissue oxygen demand and utilization and is useful when studying cellular and animal physiology.

Application

The L-Lactate Assay Kit may be used for:
  • Lactate Metabolism Research
  • Tissue Metabolism Research
  • Drug Discovery

Features and Benefits

  • Simplified Process: Experience a streamlined process with the addition of only a single working reagent, reducing complexity and saving valuable time and effort.
  • Broader Detection Range: Detect L-Lactate levels accurately across a wide range, from 1 to 50 μM, using just 50 μL of sample in a 96-well plate setup, ensuring comprehensive analysis with minimal sample requirement.
  • High-Throughput Compatibility: Effortlessly integrate our kit into high-throughput handling systems, ensuring seamless operation and maintaining efficiency and accuracy in large-scale analyses.

Suitability

The kit is suitable for the quantitative determination of L-lactate (L-lactic acid) and the evaluation of drug effects on its metabolism in serum, plasma, urine, cell culture media, and other biological samples.

Principle

The L-Lactate Assay Kit is based on the lactate dehydrogenase catalyzed oxidation of lactate, in which the formed NADH reduces a probe into a highly fluorescent product. The fluorescence intensity of this product, measured at λEx = 530 nm/λEm = 585 nm, is proportional to the lactate concentration in the sample.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

related product

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. The kit contains a probe, but no instructions in the protocol of when/how much to add. Please advise on the probe-adding step?

    1 answer
    1. The protocol indicates that 5 uL of the probe should be added to the working reagent. This is described in Table 2, located on page 3 of the datasheet. Please see the link below to review:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/429/213/mak443pis-ms.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service