Skip to Content
Merck
All Photos(3)

Key Documents

MAK062

Sigma-Aldrich

IDH Activity Assay Kit

sufficient for 100 colorimetric tests

Synonym(s):

Isocitrate Dehydrogenase Activity Assay

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$659.00

$659.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$659.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

$659.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

usage

sufficient for 100 colorimetric tests

detection method

colorimetric

relevant disease(s)

cancer

storage temp.

−20°C

General description

Isocitrate dehydrogenase (IDH) catalyzes the conversion of isocitrate to α-ketoglutarate. In eukaryotes, there are three isozymes of IDH, the mitochondrial IDH2 and IDH3, and the cytoplasmic/ peroxisomal IDH1. All three IDH family members require the presence of a divalent cation (Mg2+ or Mn2+) and either the electron-accepting cofactor NADP+ (IDH1 and IDH2) or NAD+ (IDH3) for their enzymatic activity. IDH1 and IDH2 mutations resulting in neomorphic enzymatic activity are found in certain cancers such as glioblastoma, acute myeloid leukemia, and colon cancer. This neoactivity shows a change in the substrate specificity resulting in the conversion of α-ketoglutarate to 2-hydroxyglutarate. Mutations in IDH family members are also associated with Ollier disease and Maffucci syndrome.

Application

IDH Activity Assay Kit has been used to determine the activity of isocitrate dehydrogenase in samples.[1][2]

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the measurement of isocitrate dehydrogenase (IDH) (NAD+ - dependent, NADP+ -dependent or both IDHs) activity in biological samples including tissue, cells and serum

Principle

The Isocitrate Dehydrogenase Activity Assay kit provides a simple and direct procedure for measuring NADP+-dependent, NAD+-dependent, or both NADP+ and NAD+-dependent IDH activity in a variety of samples. IDH activity is determined using isocitrate as the substrate in an enzyme reaction, which results in a colorimetric (450 nm) product proportional to the enzymatic activity present. One unit of IDH is the amount of enzyme that will generate 1.0 μmole of NADH or NADP per minute at pH 8.0 at 37 °C.

related product

Product No.
Description
Pricing

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trilobatin Protects Against Oxidative Injury in Neuronal PC12 Cells Through Regulating Mitochondrial ROS Homeostasis Mediated by AMPK/Nrf2/Sirt3 Signaling Pathway.
Gao J, et al.
Frontiers in Molecular Neuroscience, 11, 267-267 (2018)
Jianmei Gao et al.
Frontiers in molecular neuroscience, 11, 267-267 (2018-08-15)
Oxidative stress-induced neuronal cell damage is a crucial factor in the pathogenesis of mitochondria-associated neurological diseases. Therefore, elimination of overproduction of mitochondrial reactive oxygen species (mtROS) may be a potential strategy for prevention and treatment of neurological diseases. In the
Maite Garcia-Guasch et al.
The Journal of nutritional biochemistry, 99, 108833-108833 (2021-08-03)
Breast cancer is the most common malignancy in women worldwide, and environmental factors, especially diet, have a role in the etiology of this disease. This work aimed to investigate the influence of high fat diets (rich in corn oil or
Enrique Chávez et al.
The Journal of pharmacology and experimental therapeutics, 361(2), 292-302 (2017-02-18)
Background: Mitochondrion is an important metabolic and energetic organelle that regulates several cellular processes. Mitochondrial dysfunction has been related to liver diseases including hepatocellular carcinoma. As a result, the energetic demand is not properly supplied and mitochondrial morphologic changes have
Ting La et al.
Theranostics, 11(19), 9605-9622 (2021-10-15)
Rationale: Recurrent and metastatic cancers often undergo a period of dormancy, which is closely associated with cellular quiescence, a state whereby cells exit the cell cycle and are reversibly arrested in G0 phase. Curative cancer treatment thus requires therapies that

Protocols

We describe here a rapid and sensitive method to separate and measure D-2-OHG and L-2-OHG enantiomers using high-resolution mass spectrometry (HRMS) detection.

Chromatograms

application for HPLC

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service