Skip to Content
Merck
All Photos(1)

Key Documents

M1438

Sigma-Aldrich

(±)-2-Methylarachidonoyl-2′-fluoroethylamide

≥95%, ethanol solution

Synonym(s):

2-Methyl-2′-fluoro AEA, Fluoromethanandamide, O-689

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$227.00

$227.00


Estimated to ship onApril 16, 2025



Select a Size

Change View
5 MG
$227.00

About This Item

Empirical Formula (Hill Notation):
C23H38NOF
CAS Number:
Molecular Weight:
363.55
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

$227.00


Estimated to ship onApril 16, 2025


Quality Level

Assay

≥95%

form

ethanol solution

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCC(C)C(=O)NCCF

InChI

1S/C23H38FNO/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-22(2)23(26)25-21-20-24/h7-8,10-11,13-14,16-17,22H,3-6,9,12,15,18-21H2,1-2H3,(H,25,26)/b8-7-,11-10-,14-13-,17-16-

InChI key

HMMNZALKMVCHHZ-ZKWNWVNESA-N

Biochem/physiol Actions

An analog of arachidonylethanolamide (anandamide) that is a potent, selective CB1 cannabinoid receptor agonist. The methyl group at the C-2 position of arachidonic acid gives the compound enhanced metabolic stability. It can fully substitute for Δ9-THC in animal self-administration tests.
Anandamide analog; potent, selective CB1 cannabinoid receptor agonist.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service