Skip to Content
Merck
All Photos(1)

Documents

M1936

Greiner 384 well plates, polypropylene

conical bottom clear polypropylene wells

Synonym(s):

384 multiwell plates, 384 well microplates, 384 well microtiter plates, 384 well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.22

material

clear polypropylene plate
conical bottom clear polypropylene wells

sterility

non-sterile

feature

lid: no

packaging

case of 60

manufacturer/tradename

Greiner 781270

maximum volume

240 μL

size

384 wells

working volume

225 μL

color

clear

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

384 well microplate, non-treated microtiter plate with an optimal working volume of 20-225 μL; maximum well capacity of 240 μL.
Multiwell plates from Greiner. Standard microplate footprint. Excellent optical properties. DNAse/RNAse free. Non-pyrogenic by LAL method to <0.06 EU/mL.

Appropriate for applications requiring increased chemical resistance or heat stability, e.g. liquid storage or compound libraries in drug screening.

Other Notes

384 conical (V) bottom microtiter plate, without lid, rounded square well design, alphanumeric well coding; length: 127.76 +/- 0.15 mm; width: 85.48 +/- 0.15 mm; 22 mm height; curvature: approx. 200 μm.

Suitability

Free of detectable DNase/RNase, human DNA and pyrogens. Appropriate for applications requiring increased chemical resistance or heat stability, e.g. liquid storage or compound libraries in drug screening.

Legal Information


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service