Skip to Content
Merck
All Photos(1)

Key Documents

BR701355

BRAND® 384-well plate, deep well

volume 0.3 mL, polypropylene

Synonym(s):

Deep-well Plates, Sample Storage

Sign Into View Organizational & Contract Pricing

Select a Size

48 EA
$1,800.00

$1,800.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
48 EA
$1,800.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
polypropylene
size:
384 well
sterility:
non-sterile
binding type:
non-treated surface
feature:
skirt

$1,800.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

polypropylene

description

alphanumeric identification system

sterility

non-sterile

feature

skirt

packaging

pack of 48 ea

manufacturer/tradename

BRAND 701355

size

384 well

volume

0.3 mL

working volume

300 μL

binding type

non-treated surface

General description

  • BRAND® 384-well deep well plates are made of high-grade chemically resistant polypropylene.
  • Ideal for use in various automated processes including High-Throughput Screening (HTS), cell and tissue cultures, serial dilutions, reagent transfer, and sample storage down to -80 °C.
  • The raised edges of the well facilitate a secure seal and prevent sample contamination.
  • Stackable - A convenient space-saving option.
  • "V" bottom base ensures optimal sampling.
  • Alphanumeric coding for easy identification.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service