Skip to Content
Merck
All Photos(1)

Key Documents

M046030

Sigma-Aldrich

Cyanine 5 Phosphoramidite

configured for ABI ÄKTA® and OligoPilot®

Synonym(s):

2-[5-[1,3-Dihydro-1-[3-(4-methoxytriphenylmethoxy)propyl]-3,3-dimethyl-2H-indol-2-ylidene]-1,3-pentadien-1-yl]-1-[3-[N,N-diisopropylamino(2-cyanoethoxy)phosphinoxy]propyl]-3,3-dimethyl-3H-indolium

Sign Into View Organizational & Contract Pricing

Select a Size

0.05 MMOL
$734.00
0.1 MMOL
$1,270.00
0.25 G
$2,940.00

$734.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.05 MMOL
$734.00
0.1 MMOL
$1,270.00
0.25 G
$2,940.00

About This Item

Empirical Formula (Hill Notation):
C60H72N4O4P
CAS Number:
Molecular Weight:
944.21
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

$734.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

product line

Proligo Reagents

Assay

≥90% (31P-NMR)
≥95.0% (reversed phase HPLC)

form

solid

mol wt

944.21 g/mol

color

clear blue0.1M in Acetonitrile

solubility

acetonitrile: 0.1 M, clear, blue

compatibility

configured for ABI ÄKTA® and OligoPilot®

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC(C1=C(N/2CCCOP(N(C(C)C)C(C)C)OCCC#N)C=CC=C1)(C)C2=C\C=C\C=C\C(C3(C)C)=[N+](CCCOC(C4=CC=CC=C4)(C5=CC=C(OC)C=C5)C6=CC=CC=C6)C7=C3C=CC=C7.[Cl-]

InChI

1S/C60H72N4O4P.ClH/c1-46(2)64(47(3)4)69(67-44-23-40-61)68-45-25-42-63-55-33-22-20-31-53(55)59(7,8)57(63)35-18-12-17-34-56-58(5,6)52-30-19-21-32-54(52)62(56)41-24-43-66-60(48-26-13-10-14-27-48,49-28-15-11-16-29-49)50-36-38-51(65-9)39-37-50;/h10-22,26-39,46-47H,23-25,41-45H2,1-9H3;1H/q+1;/p-1

InChI key

GWCNEDQILVVBDJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Phosphoramidite technology is ideally suited for the modification ofsynthetic oligonucleotides with covalently attached reporter moieties,linkers, spacers, or haptens. Sigma-Aldrich provides popular reporters andlinkers ready to use for DNA/RNA synthesis machines such as β-cyanoethylphosphoramidites. Cyanine 5 Phosphoramidite is one such label whose absorbance maxima is 646 nm and emission maxima is 662nm.Cyanine 5 Phosphoramidite is configured for ABI Synthesizers.

Legal Information

ABI is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
OligoPilot is a registered trademark of Cytiva
ÄKTA is a registered trademark of Cytiva

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service