Skip to Content
Merck
All Photos(1)

Key Documents

M047030

Sigma-Aldrich

Cyanine 3 Phosphoramidite

configured for ABI

Synonym(s):

2-[3-[1,3-Dihydro-1-[3-(4-methoxytriphenylmethoxy)propyl]-3,3-dimethyl-2H-indol-2-ylidene]-1-propen-1-yl]-1-[3-[N,N-diisopropylamino(2-cyanoethoxy)phosphinoxy]propyl]-3,3-dimethyl-3H-indolium

Sign Into View Organizational & Contract Pricing

Select a Size

M047030-0.05MMOL
$807.00
M047030-0.1MMOL
$1,470.00
0.25 G
$3,320.00

$807.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
M047030-0.05MMOL
$807.00
M047030-0.1MMOL
$1,470.00
0.25 G
$3,320.00

About This Item

Empirical Formula (Hill Notation):
C58H70N4O4P
CAS Number:
Molecular Weight:
918.17
UNSPSC Code:
41116105
NACRES:
NA.21

$807.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

product line

Proligo Reagents

Quality Level

Assay

≥90% (31P-NMR)
≥95.0% (@549nm, reversed phase HPLC)

form

solid

mol wt

953.64 g/mol

technique(s)

DNA synthesis: suitable

color

dark red, 0.1M in Acetonitrile

turbidity

clear

solubility

acetonitrile: 0.1 M, clear, dark red

λ

conforms (UV/VIS Identity)

compatibility

configured for ABI

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

General description

Phosphoramidite technology is ideally suited for the modification ofsynthetic oligonucleotides with covalently attached reporter moieties,linkers, spacers or haptens. Sigma-Aldrich provides popular reporters andlinkers ready to use for DNA/RNA synthesis machines such as β-cyanoethylphosphoramidites.

Application

Cyanine 3 Phosphoramidite is designated for incorporation (internally) within oligonucleotides during automated DNA synthesis. It has been used in Fluorescence labeling and imaging studies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service