Skip to Content
Merck
All Photos(1)

Key Documents

M041080

Sigma-Aldrich

Fluorescein Phosphoramidite

configured for PerkinElmer, configured for Polygen

Synonym(s):

Fluorescein amidite

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 G
$326.00

$326.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.1 G
$326.00

About This Item

Empirical Formula (Hill Notation):
C46H58N3O10P
Molecular Weight:
843.94
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

$326.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥95% (31P-NMR)

suitability

conforms to structure for H-NMR

compatibility

configured for PerkinElmer
configured for Polygen

storage temp.

−20°C

SMILES string

CC(C)N(C(C)C)P(OCCCCCCNC(=O)c1ccc2c(c1)C(=O)OC23c4ccc(OC(=O)C(C)(C)C)cc4Oc5cc(OC(=O)C(C)(C)C)ccc35)OCCC#N

InChI

1S/C46H58N3O10P/c1-29(2)49(30(3)4)60(55-25-15-22-47)54-24-14-12-11-13-23-48-40(50)31-16-19-35-34(26-31)41(51)59-46(35)36-20-17-32(56-42(52)44(5,6)7)27-38(36)58-39-28-33(18-21-37(39)46)57-43(53)45(8,9)10/h16-21,26-30H,11-15,23-25H2,1-10H3,(H,48,50)

InChI key

XUQUNBOKLNVMMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda S Bruegl et al.
Cancer prevention research (Philadelphia, Pa.), 7(7), 686-697 (2014-04-29)
Clinical criteria, primarily young age of cancer onset and family history of signature cancers, have been developed to identify individuals at elevated risk for Lynch syndrome with the goals of early identification and cancer prevention. In 2007, the Society of
Martin G Belinsky et al.
BMC cancer, 15, 887-887 (2015-11-12)
Approximately 10-15 % of gastrointestinal stromal tumors (GISTs) lack gain of function mutations in the KIT and platelet-derived growth factor receptor alpha (PDGFRA) genes. An alternate mechanism of oncogenesis through loss of function of the succinate-dehydrogenase (SDH) enzyme complex has
Anna J Henningsson et al.
BMC microbiology, 15, 153-153 (2015-08-02)
A TaqMan real-time PCR assay targeting the Anaplasma citrate synthase gene, gltA, was developed and used for detection of Anaplasma phagocytophilum in 765 Ixodes ricinus ticks collected from dogs and cats in northern Norway (n = 669) and Telemark county in southern
Chiaki Taira et al.
Clinica chimica acta; international journal of clinical chemistry, 445, 101-106 (2015-03-24)
Chimerism analysis is important for the evaluation of engraftment and predicting relapse following hematopoietic stem cell transplantation (HSCT). We developed a chimerism analysis for single nucleotide polymorphisms (SNPs), including rapid screening of the discriminable donor/recipient alleles using droplet allele-specific PCR
Hiroaki Tachibana et al.
Biosensors & bioelectronics, 74, 725-730 (2015-07-27)
Polymerase chain reaction (PCR)-based genetic testing has become a routine part of clinical diagnoses and food testing. In these fields, rapid, easy-to-use, and cost-efficient PCR chips are expected to be appeared for providing such testing on-site. In this study, a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service