Skip to Content
Merck
All Photos(1)

Key Documents

L8170

Sigma-Aldrich

Pseudolaric Acid B

≥98% (HPLC)

Synonym(s):

(-)-Pseudolaric acid B, O-Acetylpseudolaric acid C, Pseudolarix acid B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H28O8
CAS Number:
Molecular Weight:
432.46
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.25

biological source

Larix kaempferi

Assay

≥98% (HPLC)

form

powder or crystals

storage condition

protect from light

color

white to off-white

solubility

methanol: 1 mg/mL, clear, colorless

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

O=C(OC)C1=CC[C@@]2(C(O[C@@](/C=C/C=C(C(O)=O)\C)(C)[C@@H]3CC2)=O)[C@]3(OC(C)=O)CC1

InChI

1S/C23H28O8/c1-14(17(24)25)6-5-10-21(2)16-9-12-22(19(27)31-21)11-7-15(18(26)29-3)8-13-23(16,22)20(28)30-4/h5-7,10,16H,8-9,11-13H2,1-4H3,(H,24,25)/b10-5+,14-6+/t16-,21+,22+,23+/m0/s1

InChI key

XRLYZNSOXNPKOR-CBDALDGHSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Pseudolaric acid B, a natural diterpenoid compound, is isolated from Pseudolarix kaempferi. It has shown antifungal, antifertility, and antiangiogenic properties. It also may exihibit anti-cancer and anti-inflammatory properties.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Taradas Sarkar et al.
Biochemical pharmacology, 84(4), 444-450 (2012-05-29)
We purified pseudolaric acid B (PAB) from the root and stem bark of Pseudolarix kaempferi (Lindl.) Gorden. Confirming previous findings, we found that the compound had high nanomolar IC₅₀ antiproliferative effects in several cultured cell lines, causing mitotic arrest and
Ai-guo Meng et al.
Acta pharmacologica Sinica, 30(4), 442-450 (2009-03-24)
The aim of this study was to investigate the mechanism of pseudolaric acid B (PLAB)-induced cell cycle arrest in human melanoma SK-28 cells. Cell growth inhibition was detected by MTT assay, the cell cycle was analyzed by flow cytometry, and
Min Qi et al.
Journal of pharmacological sciences, 121(3), 200-211 (2013-02-27)
Pseudolaric acid B (PAB) is the primary biologically active compound isolated from the root bark of P. kaempferi Gordon. Our previous study demonstrated that PAB induced mitotic catastrophe in L929 cells and indicated that only a small percentage (12%) of
Min Qi et al.
European journal of pharmacology, 683(1-3), 16-26 (2012-03-15)
Pseudolaric acid B (PAB) is the primary biologically active compound isolated from the root bark of P. kaempferi Gordon. Previous studies have demonstrated that PAB arrests cells in G2/M phase in several cancer cell lines without significantly perturbing the G2/M
Quan-Bin Han et al.
Journal of separation science, 32(2), 309-313 (2008-12-17)
In order to provide the chemical markers for the quality control of herbal medicines, four diterpenoids, pseudolaric acids A and B (PAA and PAB), and their glucosides were isolated from the methanol extract of the Chinese herb Pseudolarix kaempferi using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service