Skip to Content
Merck
All Photos(1)

Key Documents

PHL80523

Pseudolaric Acid B

phyproof® Reference Substance

Synonym(s):

(-)-Pseudolaric acid B, O-Acetylpseudolaric acid C, Pseudolarix acid B

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$584.00

$584.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$584.00

About This Item

Empirical Formula (Hill Notation):
C23H28O8
CAS Number:
Molecular Weight:
432.46
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$584.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

storage temp.

2-8°C

SMILES string

O=C(OC)C1=CC[C@@]2(C(O[C@@](/C=C/C=C(C(O)=O)\C)(C)[C@@H]3CC2)=O)[C@]3(OC(C)=O)CC1

InChI

1S/C23H28O8/c1-14(17(24)25)6-5-10-21(2)16-9-12-22(19(27)31-21)11-7-15(18(26)29-3)8-13-23(16,22)20(28)30-4/h5-7,10,16H,8-9,11-13H2,1-4H3,(H,24,25)/b10-5+,14-6+/t16-,21+,22+,23+/m0/s1

InChI key

XRLYZNSOXNPKOR-CBDALDGHSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Pseudolaric acid B, a natural diterpenoid compound, is isolated from Pseudolarix kaempferi. It has shown antifungal, antifertility, and antiangiogenic properties. It also may exihibit anti-cancer and anti-inflammatory properties.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service