Skip to Content
Merck
All Photos(1)

Key Documents

74437

Sigma-Aldrich

(+)-Nootkatone

≥99.0% (GC)

Synonym(s):

(4R,4aS,6R)-4,4a,5,6,7,8-Hexahydro-4,4a-dimethyl-6-(1-methylethenyl)-2(3H)-naphthalenone

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$207.00
2.5 G
$493.00

$207.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$207.00
2.5 G
$493.00

About This Item

Empirical Formula (Hill Notation):
C15H22O
CAS Number:
Molecular Weight:
218.33
Beilstein:
4676969
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.22

$207.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥99.0% (GC)

form

crystals

optical activity

[α]20/D +182.0±5.0°, c = 1% in ethanol

storage condition

under inert gas (Argon)

mp

35-39 °C

functional group

ketone

storage temp.

2-8°C

SMILES string

C[C@@H]1CC(=O)C=C2CC[C@H](C[C@@]12C)C(C)=C

InChI

1S/C15H22O/c1-10(2)12-5-6-13-8-14(16)7-11(3)15(13,4)9-12/h8,11-12H,1,5-7,9H2,2-4H3/t11-,12-,15+/m1/s1

InChI key

WTOYNNBCKUYIKC-JMSVASOKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(+)-nootkatone, a bicyclic conjugated sesquiterpene ketone[1] with a grapefruit-like flavor, is commonly used in fragrance, food, cosmetics and pharmaceutical industry. It can be synthesized from (+)-valencene via biotransformation.[2] (+)-nootkatone is the active ingredient responsible for the antiplatelet effect of Cyperus rotundus, a well-known oriental traditional medicine.[3] It also shows promising efficacy against Staphylococcus aureus biofilms.[4]

Application

(+)-Nootkatone can be used:
  • In the iron(III) porphyrin catalyzed alkene aziridination reaction with organic azides under photo-irradiation condition.[5]
  • As a substrate to synthesize its corresponding triene derivative through reduction-elimination reaction using iridium catalyst.[6]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iron porphyrin catalysed light driven C-H bond amination and alkene aziridination with organic azides
Du Y, et al.
Chemical Science, 11(18), 4680-4686 (2020)
Iridium-catalysed highly selective reduction--elimination of steroidal 4-en-3-ones to 3, 5-dienes in water
Li J, et al.
Green Chemistry, 21(8), 2088-2094 (2019)
Inhibition of multidrug-resistant foodborne Staphylococcus aureus biofilms by a natural terpenoid (+)-nootkatone and related molecular mechanism
Farha AK, et al.
Food Control, 112(18), 107154-107154 (2020)
Rick G Kelsey et al.
Journal of chemical ecology, 31(11), 2653-2670 (2005-11-08)
We measured the concentrations of extractable bioactive compounds in heartwood of live yellow-cedar (Chamaecyparis nootkatensis) trees and five classes of standing snags (1-5, averaging 4, 14, 26, 51, and 81 years-since-death, respectively) to determine how the concentrations changed in the
Marc C Dolan et al.
Journal of economic entomology, 102(6), 2316-2324 (2010-01-15)
We evaluated the ability of the natural, plant-derived acaricides nootkatone and carvacrol to suppress Ixodes scapularis Say and Amblyomma americanum (L.) (Acari: Ixodidae). Aqueous formulations of 1 and 5% nootkatone applied by backpack sprayer to the forest litter layer completely

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service