Skip to Content
Merck
All Photos(1)

Documents

L7670

Sigma-Aldrich

PL405 trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

AcSKGPya-F-W-Nop-GGKNH2 trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C71H83N15O14 · xC2HF3O2
CAS Number:
Molecular Weight:
1370.51 (free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

acetonitrile: water (1:1): ≥10 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CC(=O)N[C@@H](CO)C(=O)N[C@@H](CCCCN)C(=O)NCC(=O)N[C@@H](Cc1cc2cccc3ccc4cccc1c4c23)C(=O)N[C@@H](Cc5ccccc5)C(=O)N[C@@H](Cc6c[nH]c7ccccc67)C(=O)N[C@@H](Cc8ccc(cc8)[N+]([O-])=O)C(=O)NCC(=O)NCC(=O)N[C@@H](CCCCN)C(N)=O

InChI

1S/C71H83N15O14.C2HF3O2/c1-41(88)79-59(40-87)71(98)82-54(22-8-10-30-73)66(93)78-39-62(91)81-57(34-47-33-46-17-11-15-44-25-26-45-16-12-19-51(47)64(45)63(44)46)69(96)84-56(31-42-13-3-2-4-14-42)68(95)85-58(35-48-36-75-52-20-6-5-18-50(48)52)70(97)83-55(32-43-23-27-49(28-24-43)86(99)100)67(94)77-37-60(89)76-38-61(90)80-53(65(74)92)21-7-9-29-72;3-2(4,5)1(6)7/h2-6,11-20,23-28,33,36,53-59,75,87H,7-10,21-22,29-32,34-35,37-40,72-73H2,1H3,(H2,74,92)(H,76,89)(H,77,94)(H,78,93)(H,79,88)(H,80,90)(H,81,91)(H,82,98)(H,83,97)(H,84,96)(H,85,95);(H,6,7)/t53-,54-,55-,56-,57-,58-,59-;/m0./s1

InChI key

NVRXYBFFJSLZOM-DMJUDXNFSA-N

Biochem/physiol Actions

PL405 is a specific and high affinity endothelin converting enzyme-2 (ECE-2) fluorigenic substrate. PL405 is cleaved by ECE-2 at the Pya-F amide bond to yield the highly fluorigenic Ac-SKG-Pya.

Features and Benefits

This compound is featured on the Endothelin Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service