Skip to Content
Merck
All Photos(4)

Documents

L2646

Sigma-Aldrich

Lectin from Phaseolus vulgaris (red kidney bean)

Phytohemagglutinin PHA-M, lyophilized powder

Synonym(s):

Phaseolus vulgaris agglutinin, PHA

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352202
NACRES:
NA.32

Quality Level

form

lyophilized powder

potency

<40 μg per mL agglutination activity

composition

Protein, 50-70% biuret

impurities

salt, free

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

PHA consists of two molecular species, an erythroagglutinin (PHA-E) which has low mitogenic activity and high erythroagglutinin activity, and leucoagglutinin (PHA-L) which has high mitogenic and leucoagglutinating activity, but very low erythroagglutinating activity.
PHA-E is not blood group specific, but agglutination can be inhibited by certain oligosaccharides. PHA-P is the protein form of PHA prior to separation and purification of erythroagglutinin and leucoagglutinin. PHA-M is the mucoprotein form. Conjugates are prepared from the corresponding purified lectins.

Analysis Note

Agglutination activity is expressed in μg/mL and is determined from serial dilutions of a 1 mg/mL solution. This activity is the lowest concentration to agglutinate a 2% suspension of human erythrocytes in phosphate buffered saline, pH 6.8, after 1 hr incubation at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaohua Chen et al.
PLoS neglected tropical diseases, 13(4), e0007318-e0007318 (2019-04-25)
Leprosy, caused by Mycobacterium leprae, affects over 200,000 people annually worldwide and remains endemic in the ethnically diverse, mountainous and underdeveloped southwestern provinces of China. Delayed diagnosis of leprosy persists in China, thus, additional knowledge to support early diagnosis, especially
Balam Ruiz-Ruiz et al.
ACS omega, 5(21), 12005-12015 (2020-06-18)
Silver nanoparticles (AgNPs) are the most used nanomaterials worldwide due to their excellent antibacterial, antiviral, and antitumor activities, among others. However, there is scarce information regarding their genotoxic potential measured using human peripheral blood lymphocytes. In this work, we present
Lucía Margarita Valenzuela-Salas et al.
Pharmaceutics, 13(1) (2021-01-13)
Nanomaterials quickly evolve to produce safe and effective biomedical alternatives, mainly silver nanoparticles (AgNPs). The AgNPs' antibacterial, antiviral, and antitumor properties convert them into a recurrent scaffold to produce new treatment options. This work reported the full characterization of a
Jisun Lim et al.
Science advances, 6(16), eaba1334-eaba1334 (2020-06-04)
Glutathione (GSH), the most abundant nonprotein thiol functioning as an antioxidant, plays critical roles in maintaining the core functions of mesenchymal stem cells (MSCs), which are used as a cellular immunotherapy for graft-versus-host disease (GVHD). However, the role of GSH

Articles

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service