Skip to Content
Merck
All Photos(1)

Documents

C2743

Sigma-Aldrich

Cytochalasin B from Drechslera dematioidea

Ready Made Solution, 10 mg/mL in DMSO

Synonym(s):

Phomin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H37NO5
CAS Number:
Molecular Weight:
479.61
Beilstein:
1096207
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

Quality Level

form

solution

concentration

10 mg/mL in DMSO

antibiotic activity spectrum

fungi

Mode of action

cell membrane | interferes

storage temp.

−20°C

SMILES string

C[C@@H]1CCC[C@@H](O)\C=C\C(=O)O[C@]23[C@@H](\C=C\C1)[C@H](O)C(=C)[C@@H](C)[C@H]2[C@H](Cc4ccccc4)NC3=O

InChI

1S/C29H37NO5/c1-18-9-7-13-22(31)15-16-25(32)35-29-23(14-8-10-18)27(33)20(3)19(2)26(29)24(30-28(29)34)17-21-11-5-4-6-12-21/h4-6,8,11-12,14-16,18-19,22-24,26-27,31,33H,3,7,9-10,13,17H2,1-2H3,(H,30,34)/b14-8+,16-15+/t18-,19-,22-,23+,24+,26+,27-,29-/m1/s1

InChI key

GBOGMAARMMDZGR-TYHYBEHESA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Cytochalasin B from Drechslera dematioidea has been used:
  • to block cytokinesis, to obtain binucleated cells
  • as a specific glucose transporter inhibitor
  • to inhibit glucose transport by class I and III GLUTs to determine the 50% inhibitory concentration (IC50) in HCMV-infected and mock-infected cells

Cytochalasin B is used to inhibit sugar transport in fibroblasts , decrease cytoskeletal actin and myosin in rabbit neutrophils , and to stimulate B lymphocytes .

Biochem/physiol Actions

Cell permeable fungal toxin that disrupts contractile microfilaments by inhibiting actin polymerization. This, in turn, induces DNA fragmentation, inhibits cell division, and disrupts many cell processes. Inhibits glucose transport.
Cytochalasin B can prevent cytoplasmic division without affecting nuclear division.
One of a group of fungal metabolites that interfere with a wide variety of cellular movements. Useful tool for characterizing some of the polymerization properties of actin, and in studies on cytokinesis. Probe for the two hexose-transport systems in rat L6 myoblasts.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

185.0 °F - closed cup - Solvent

Flash Point(C)

85 °C - closed cup - Solvent

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael V Zuccaro et al.
Cell, 183(6), 1650-1664 (2020-10-31)
Correction of disease-causing mutations in human embryos holds the potential to reduce the burden of inherited genetic disorders and improve fertility treatments for couples with disease-causing mutations in lieu of embryo selection. Here, we evaluate repair outcomes of a Cas9-induced
Adam R Akkad et al.
PloS one, 17(5), e0268508-e0268508 (2022-05-21)
Biological materials can be shipped off-site for diagnostic, therapeutic and research purposes. They usually are kept in certain environments for their final application during transportation. However, active reagent handling during transportation from a collection site to a laboratory or biorepository
[18 F] FDG PET signal is driven by astroglial glutamate transport
Zimmer ER, et al.
Nature Neuroscience, 20(3), 393-393 (2017)
Alternative In Vitro Models for Safety and Toxicity Evaluation of Nutraceuticals
Nutraceuticals, 355-385 (2016)
Human Cytomegalovirus Activates Glucose Transporter 4 Expression To Increase Glucose Uptake during Infection
Yu Y, et al.
Journal of virology, 85(4), 1573-1580 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service