Skip to Content
Merck
All Photos(1)

Key Documents

PZ0266

Sigma-Aldrich

PHA-408

≥98% (HPLC)

Synonym(s):

8-(5-Chloro-2-(4-methylpiperazin-1-yl)isonicotinamido)-1-(4-fluorophenyl)-4,5,dihydro-1H-benzo[g]indazole-3-carboxamide, PHA-00767408, PHA-767408

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$144.00
25 MG
$580.00

$144.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$144.00
25 MG
$580.00

About This Item

Empirical Formula (Hill Notation):
C29H27ClFN7O2
CAS Number:
Molecular Weight:
560.02
UNSPSC Code:
12352200
NACRES:
NA.77

$144.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

shipped in

dry ice

storage temp.

−20°C

SMILES string

Fc1ccc(cc1)[n]2nc(c3c2c4c(ccc(c4)NC(=O)c5c(cnc(c5)N6CCN(CC6)C)Cl)CC3)C(=O)N

InChI

1S/C29H27ClFN7O2/c1-36-10-12-37(13-11-36)25-15-23(24(30)16-33-25)29(40)34-19-6-2-17-3-9-21-26(28(32)39)35-38(27(21)22(17)14-19)20-7-4-18(31)5-8-20/h2,4-8,14-16H,3,9-13H2,1H3,(H2,32,39)(H,34,40)

InChI key

ZLEZHGHFWIHCGU-UHFFFAOYSA-N

Biochem/physiol Actions

PHA-408 is a potent, selective I kappa B-kinase 2 (IKK-2) inhibitor. PHA-408 is a tight binding, ATP competitive IKK-2 inhibitor with greater than 350-fold selectivity for IKK-2 vs IKK-1 (IC50 values of 10-40 nM at IKK-2 vs 14 μM at IKK-1). PHA-408 was shown to suppress inflammatory cellular events, and to reduce inflammation in animal models of arthritis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Zhong et al.
American journal of physiology. Lung cellular and molecular physiology (2020-05-07)
Cyclic guanosine monophosphate (cGMP) signaling is an important regulator of newborn lung function and development. Although cGMP signaling is decreased in many models of newborn lung injury, the mechanisms are poorly understood. We determined how IL-1β regulates the expression of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service