Key Documents
HS120116
Nitrogen-free weighing papers
W × L 3 × 3 in.
About This Item
UNSPSC Code:
41121812
NACRES:
NB.35
Pricing and availability is not currently available.
manufacturer/tradename
Heathrow Scientific HS120116
W × L
3 × 3 in.
color
white
General description
Nitrogen-free for a very smooth surface and good sample recovery. Strong enough to support dense material.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Questions
  1. What is the thickness of this butter paper?
    1 answer
    1. The thickness of this weighing paper is 0.0015” or 0.038 mm.
      Helpful?