Key Documents
HS120117
Nitrogen-free weighing papers
W × L 4 × 4 in.
Select a Size
500 EACH
$140.00
$140.00
Estimated to ship onMay 23, 2025
Select a Size
Change View
500 EACH
$140.00
About This Item
UNSPSC Code:
41121812
NACRES:
NB.35
$140.00
Estimated to ship onMay 23, 2025
manufacturer/tradename
Heathrow Scientific HS120117
W × L
4 × 4 in.
color
white
General description
Nitrogen-free for a very smooth surface and good sample recovery. Strong enough to support dense material.
Choose from one of the most recent versions:
Certificates of Analysis (COA)