Skip to Content
Merck
All Photos(3)

Documents

HPA018903

Sigma-Aldrich

Anti-GRK6 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-G protein-coupled receptor kinase 6, Anti-G protein-coupled receptor kinase GRK6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

LLFREFCATRPELSRCVAFLDGVAEYEVTPDDKRKACGRQLTQNFLSHTGPDLIPEVPRQLVTNCTQRLEQGPCKDLFQELTRLTHEYLSVAPFADYLDSIYFNRFLQWKWLER

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GRK6(2870)

General description

The gene GRK6 (G protein-coupled receptor kinase 6) is mapped to human chromosome 5q35. GRK6 transcript is expressed in various tissues including heart, brain, placenta, lung, liver, kidney, pancreas and skeletal muscle. It is a membrane associated protein.

Immunogen

G protein-coupled receptor kinase 6 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GRKs (G protein-coupled receptor kinases) generally phosphorylate agonist-activated G-protein coupled receptors and cause desensitization of these receptors. GRK6 phosphorylate chemokine receptor CXCR4 (C-X-C chemokine receptor type 4) and negatively regulate its signaling. GRK6 binds and phosphorylates low density lipoprotein receptor-related proteins-6 (LRP6), thereby resulting in LRP6 activation and Wnt/LRP6 signaling. GRK6 levels are reduced in elderly patients with schizophrenia. It is up-regulated in hepatocellular carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73516

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E R Bychkov et al.
Neurobiology of disease, 44(2), 248-258 (2011-07-26)
Alterations of multiple G protein-mediated signaling pathways are detected in schizophrenia. G protein-coupled receptor kinases (GRKs) and arrestins terminate signaling by G protein-coupled receptors exerting a powerful influence on receptor functions. Modifications of arrestin and/or GRKs expression may contribute to
John M Busillo et al.
The Journal of biological chemistry, 285(10), 7805-7817 (2010-01-06)
The chemokine receptor CXCR4 is a widely expressed G protein-coupled receptor that has been implicated in a number of diseases including human immunodeficiency virus, cancer, and WHIM syndrome, with the latter two involving dysregulation of CXCR4 signaling. To better understand
F Bullrich et al.
Cytogenetics and cell genetics, 70(3-4), 250-254 (1995-01-01)
G protein-coupled receptor kinases (GRKs) play an important role in phosphorylating and regulating the activity of a variety of G protein-coupled receptors. Chromosomal mapping of the human genes for the two most recently identified members of the GRK family, GRK5
Ya-Ping Li
Asian Pacific journal of tropical medicine, 6(3), 220-223 (2013-02-05)
To investigate the expression and potential roles of G protein-coupled receptor kinase 6 (GRK6) in hepatocellular carcinoma (HCC) patients. Immunohistochemistry and Western blot was performed to determine GRK6 expression in 73 HCC samples. And the correlation with clinicopathological features was
Minyong Chen et al.
The Journal of biological chemistry, 284(50), 35040-35048 (2009-10-06)
Wnt ligands conduct their functions in canonical Wnt signaling by binding to two receptors, the single transmembrane low density lipoprotein receptor-related proteins 5 and 6 (LRP5/6) and seven transmembrane (7TM) Frizzled receptors. Subsequently, phosphorylation of serine/threonine residues within five repeating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service