Skip to Content
Merck
All Photos(1)

Documents

GE29401324

Sigma-Aldrich

Strep-Tactin XT Sepharose chromatography resin

pkg of 1 × 10 mL, chromatography resin

Synonym(s):

StrepTrap High Performance

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
SB.53

grade

for chromatography

packaging

pkg of 1 × 10 mL

manufacturer/tradename

Cytiva 29401324

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Strep-Tactin XT Sepharose chromatography resin is ideal for the purification of recombinant proteins tagged with Strep-tag II and Twin-Strep-tag. These proteins bind very specifically to the immobilized Strep-Tactin XT ligand giving a highly pure target protein. Strep-Tactin XT Sepharose contains the latest Strep-Tactin ligand, XT, which provides higher affinity, resulting in higher yields and lower loss of valuable target protein than with the predecessor product, StrepTactin Sepharose High Performance (HP). Strep-Tactin XT Sepharose also enables purification under denaturing conditions.

Application

Strep-Tactin XT Sepharose chromatography resin is suitable for the purification of recombinant proteins tagged with a Strep-tag& II and Twin-Strep-tag.

Features and Benefits

  • Ensures highly pure Strep-tag II and Twin-Strep-tag recombinant proteins
  • Fast purifications with a simple elution process
  • Physiological conditions and mild elution preserve target protein activity
  • Fast and easy regeneration of the purification system
  • Compatible with a range of detergents, denaturants, and other additives, allowing flexibility in experimental conditions
  • Stable in commonly used aqueous buffers, ensuring reliable performance across different experiments

Legal Information

StrepTrap is a trademark of Cytiva

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service