Merck
  • Chromosomal mapping of the genes GPRK5 and GPRK6 encoding G protein-coupled receptor kinases GRK5 and GRK6.
Chromosomal mapping of the genes GPRK5 and GPRK6 encoding G protein-coupled receptor kinases GRK5 and GRK6.
Cytogenetics and cell genetics (1995-01-01)
F Bullrich, T Druck, P Kunapuli, J Gomez, K W Gripp, B Schlegelberger, J Lasota, M Aronson, L A Cannizzaro, K Huebner
ABSTRACT
G protein-coupled receptor kinases (GRKs) play an important role in phosphorylating and regulating the activity of a variety of G protein-coupled receptors. Chromosomal mapping of the human genes for the two most recently identified members of the GRK family, GRK5 (GPRK5) and GRK6 (GPRK6), was accomplished by correlation of the presence of the GPRK5 and GPRK6 loci with specific chromosome regions in a rodent-human hybrid panel. These analyses revealed that GPRK5 maps to chromosome region 10q24-->qter while GPRK6 maps to 5q35. A GPRK6 related locus maps to 13pter-->q21.