Skip to Content
Merck
All Photos(1)

Key Documents

H8765

Sigma-Aldrich

Anti-Human Serum antibody produced in rabbit

whole antiserum

Synonym(s):

Rabbit Anti-Human Antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

human

technique(s)

immunoelectrophoresis: suitable

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Serum is the component of blood plasma without cells and clotting factors. It is used in various assays, research and also as a supplement in cell culture. Anti-human serum antibody can be used in immunoelectrophoresis. Rabbit anti-human serum antibody reacts specifically with normal human serum.

Immunogen

Pooled normal human serum.

Application

Anti-human serum antibody (diluted 1:3 in KnockOut Dulbeccos′s modified Eagles medium) can be used in immunosurgery.

Quality

The antiserum has been treated to remove lipoproteins.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regional differences in expression of specific markers for human embryonic stem cells
Laursen SB, et al.
Reproductive Biomedicine Online, 15(1) (2007)
Evaluation of 28 human embryonic stem cell lines for use as unrelated donors in stem cell therapy: implications of HLA and ABO genotypes
Lee JE, et al.
Cell Transplantation, 19(11) (2010)
From Pluripotency to Early Differentiation of Human Embryonic Stem Cell Cultures Evaluated by Electron Microscopy and Immunohistochemistry.
Janus Valentin Jacobsen
Embryonic Stem Cells - Basic Biology to Bioengineering, doi: 10-doi: 10 (2011)
Christian B Brøchner et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 60(3), 188-204 (2011-12-06)
The secreted glycoprotein YKL-40 participates in cell differentiation, inflammation, and cancer progression. High YKL-40 expression is reported during early human development, but its functions are unknown. Six human embryonic stem cell (hESC) lines were cultured in an atmosphere of low

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service