234395
Complement, Guinea Pig Serum
Complement, Guinea Pig Serum, is suitable for evaluating functional activity of human complement components and for other research requiring a high level of hemolytic activity.
Synonym(s):
Complement Serum, Guinea Pig Serum
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
form
lyophilized solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
shipped in
wet ice
storage temp.
−20°C
General description
Complement, Guinea pig serum is suitable for evaluating the functional activity of human complement components and for other research requiring a high level of hemolytic activity.
Application
Complement, Guinea pig serum has been used:
- as a component of M2 medium for differential staining of blastocysts
- along with anti-human whole serum antibody to remove trophectoderm (TE) by immunosurgery
- in influenza microneutralization assay and complement-dependent cytotoxicity (CDC) assay
Biochem/physiol Actions
Functionally equivalent to reference standard 71383
Warning
Toxicity: Standard Handling (A)
Reconstitution
Following reconstitution and sterile filtration, aliquot, and freeze (-70°C). Stock solutions are stable for up to 1 year at -70°C.
Reconstitute with 5 ml of sterile, distilled H₂O. It is recommended that the reconstituted material be filter sterilized.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Iranian journal of reproductive medicine, 10(6), 555-560 (2012-11-01)
Indisputable population exposure to widespread electromagnetic fields, has grown concerns over the probable health effects of these fields. The present study was aimed to examine the possible effects of 50 Hz extremely low frequency electromagnetic field (ELF-EMF) exposure on the
Cell transplantation, 19(11), 1383-1395 (2010-07-01)
For human embryonic stem cells (hESCs) to be used clinically, it is imperative that immune responses evoked by hESCs and their derivates after transplantation should be prevented. Human leukocyte antigens (HLA) and ABO blood group antigens are important histocompatibility factors
Journal of the Chemical Society. Perkin Transactions 1, 2979-2979 (1993)
Cell, 177(5), 1124-1135 (2019-05-18)
Vaccines to generate durable humoral immunity against antigenically evolving pathogens such as the influenza virus must elicit antibodies that recognize conserved epitopes. Analysis of single memory B cells from immunized human donors has led us to characterize a previously unrecognized
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service