Skip to Content
Merck
All Photos(1)

Key Documents

H9640

Sigma-Aldrich

Anti-Human Serum antibody produced in goat

whole antiserum

Synonym(s):

Anti-Human Serum

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
$229.00

$229.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2 ML
$229.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

$229.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

goat

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

human

technique(s)

immunoelectrophoresis: suitable

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Human serum albumin consists of three homologous domains that gather to form a heart-shaped molecule. Each domain is a product of two subdomains that hold collective structural motifs.
Serum is the liquid component of blood that includes proteins, immunoglobulin and electrolytes. It has a crucial role in several diagnostic tests. Anti-human serum antibody can be used in immunoelectrophoresis. Goat anti-human serum antibody reacts specifically with normal human serum.

Immunogen

Pooled normal human serum.

Application

Anti-human serum antibody can be used in immunosurgery.

Physical form

Goat Anti-Human Serum is provided as a liquid containing 0.1% sodium azide as preservative

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teija Peura et al.
Current protocols in stem cell biology, Chapter 1, Unit1A-Unit1A (2011-03-15)
This unit describes generation of human embryonic stem cell lines from early human embryos. The focus is on actual handling of embryos and early embryonic outgrowths, omitting steps required for actual generation, freezing, and thawing of embryos, as well as
Atomic structure and chemistry of human serum albumin
He X M and Carter D C
Nature, 358(6383), 209-209 (1992)
B E Reubinoff et al.
Nature biotechnology, 18(4), 399-404 (2000-04-05)
We describe the derivation of pluripotent embryonic stem (ES) cells from human blastocysts. Two diploid ES cell lines have been cultivated in vitro for extended periods while maintaining expression of markers characteristic of pluripotent primate cells. Human ES cells express

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service